Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 48738863-48739112 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CEP152
Entrez Gene ID
22995
Gene Name
centrosomal protein 152
Gene Aliases
MCPH4, MCPH9, SCKL5
UniGene
Hs.443005
Gene Chromosome Location
Chr.15: 48662534-48811901 on Build GRCh38
Transcripts
NM_001194998.1 NM_014985.3 XM_006720437.3 XM_011521373.2 XM_011521374.2 XM_011521375.2 XM_011521378.2 XM_011521379.2 XM_011521381.2 XM_017022014.1 XM_017022015.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3956691 c.4252G>T p.G1418C Chr.15: 48738962-48738962 on Build GRCh38
7261334 c.4505T>C p.L1502S Chr.15: 48738877-48738877 on Build GRCh38
962409 c.4301T>G p.L1434R Chr.15: 48738913-48738913 on Build GRCh38
6219819 c.4273delC p.L1425fs*22 Chr.15: 48738941-48738941 on Build GRCh38
5748499 c.4298C>G p.S1433* Chr.15: 48738916-48738916 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs754324885 Chr.15: 48739006-48739014 on Build GRCh38
rs377685429 Chr.15: 48738962-48738962 on Build GRCh38
rs375493219 Chr.15: 48739013-48739013 on Build GRCh38
rs767421643 Chr.15: 48739060-48739060 on Build GRCh38
rs757819368 Chr.15: 48739008-48739009 on Build GRCh38
rs377560589 Chr.15: 48739014-48739014 on Build GRCh38
rs777011470 Chr.15: 48739017-48739017 on Build GRCh38
rs756468243 Chr.15: 48738967-48738968 on Build GRCh38
rs747401375 Chr.15: 48739006-48739006 on Build GRCh38
rs370649310 Chr.15: 48739033-48739033 on Build GRCh38
rs781013518 Chr.15: 48738935-48738935 on Build GRCh38
rs997214199 Chr.15: 48738932-48738932 on Build GRCh38
rs764880513 Chr.15: 48738910-48738910 on Build GRCh38
rs764714488 Chr.15: 48739026-48739026 on Build GRCh38
rs761234003 Chr.15: 48738893-48738893 on Build GRCh38
rs374524698 Chr.15: 48739064-48739064 on Build GRCh38
rs367643016 Chr.15: 48739054-48739054 on Build GRCh38
rs755014174 Chr.15: 48738927-48738927 on Build GRCh38
rs745375941 Chr.15: 48739086-48739086 on Build GRCh38
rs758255290 Chr.15: 48738913-48738913 on Build GRCh38
rs114085678 Chr.15: 48739083-48739083 on Build GRCh38
rs746142826 Chr.15: 48738968-48738968 on Build GRCh38
rs765791978 Chr.15: 48738951-48738951 on Build GRCh38
rs376905337 Chr.15: 48738926-48738926 on Build GRCh38
rs1031224468 Chr.15: 48739000-48739000 on Build GRCh38
rs768919003 Chr.15: 48739086-48739086 on Build GRCh38
rs778390919 Chr.15: 48739033-48739034 on Build GRCh38
rs372379014 Chr.15: 48739062-48739062 on Build GRCh38
rs1018761537 Chr.15: 48739016-48739016 on Build GRCh38
rs372672217 Chr.15: 48738909-48738909 on Build GRCh38
rs776161221 Chr.15: 48738906-48738906 on Build GRCh38
rs200552807 Chr.15: 48738925-48738925 on Build GRCh38
rs373725563 Chr.15: 48739052-48739052 on Build GRCh38
rs533145180 Chr.15: 48738923-48738923 on Build GRCh38
rs370000548 Chr.15: 48738957-48738957 on Build GRCh38
rs141600901 Chr.15: 48739003-48739004 on Build GRCh38
rs772417645 Chr.15: 48738970-48738970 on Build GRCh38
rs780290677 Chr.15: 48739071-48739071 on Build GRCh38
rs763494656 Chr.15: 48738898-48738898 on Build GRCh38
rs753060109 Chr.15: 48738949-48738949 on Build GRCh38
rs370946718 Chr.15: 48738978-48738978 on Build GRCh38
rs781716447 Chr.15: 48739087-48739087 on Build GRCh38
rs797045455 Chr.15: 48739006-48739017 on Build GRCh38
rs951374273 Chr.15: 48739061-48739061 on Build GRCh38
rs373848269 Chr.15: 48738961-48738961 on Build GRCh38

View Full Product Details