Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 152607292-152607547 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMNL2
Entrez Gene ID
114793
Gene Name
formin like 2
Gene Aliases
FHOD2
UniGene
Hs.654630
Gene Chromosome Location
Chr.2: 152335170-152649834 on Build GRCh38
Transcripts
NM_052905.3 XM_005246263.2 XM_005246265.3 XM_011510530.1 XM_011510531.1 XM_011510532.2 XM_011510533.2 XM_011510534.1 XM_011510535.2 XM_011510536.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3406943 c.883G>A p.G295R Chr.2: 152607345-152607345 on Build GRCh38
6056025 c.895C>T p.R299C Chr.2: 152607357-152607357 on Build GRCh38
3694915 c.923G>A p.R308K Chr.2: 152607385-152607385 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs371539614 Chr.2: 152607339-152607339 on Build GRCh38
rs879190466 Chr.2: 152607472-152607473 on Build GRCh38
rs760397209 Chr.2: 152607457-152607458 on Build GRCh38
rs767425166 Chr.2: 152607376-152607376 on Build GRCh38
rs2118376 Chr.2: 152607507-152607507 on Build GRCh38
rs3080597 Chr.2: 152607495-152607496 on Build GRCh38
rs760606592 Chr.2: 152607395-152607395 on Build GRCh38
rs72210499 Chr.2: 152607471-152607474 on Build GRCh38
rs373664102 Chr.2: 152607433-152607433 on Build GRCh38
rs762989414 Chr.2: 152607373-152607373 on Build GRCh38
rs199619989 Chr.2: 152607504-152607504 on Build GRCh38
rs567281941 Chr.2: 152607384-152607384 on Build GRCh38
rs201721916 Chr.2: 152607357-152607357 on Build GRCh38
rs778187313 Chr.2: 152607497-152607499 on Build GRCh38
rs775319807 Chr.2: 152607458-152607458 on Build GRCh38
rs749937911 Chr.2: 152607426-152607426 on Build GRCh38
rs751096540 Chr.2: 152607499-152607500 on Build GRCh38
rs758876928 Chr.2: 152607473-152607474 on Build GRCh38
rs35268946 Chr.2: 152607464-152607464 on Build GRCh38
rs3080598 Chr.2: 152607471-152607472 on Build GRCh38
rs201558427 Chr.2: 152607494-152607495 on Build GRCh38
rs370334407 Chr.2: 152607401-152607401 on Build GRCh38
rs58236123 Chr.2: 152607504-152607507 on Build GRCh38
rs771648047 Chr.2: 152607327-152607328 on Build GRCh38
rs183182592 Chr.2: 152607506-152607506 on Build GRCh38
rs58684133 Chr.2: 152607506-152607507 on Build GRCh38
rs796288865 Chr.2: 152607504-152607506 on Build GRCh38
rs755563247 Chr.2: 152607444-152607444 on Build GRCh38
rs779716382 Chr.2: 152607434-152607434 on Build GRCh38
rs923125423 Chr.2: 152607506-152607507 on Build GRCh38
rs763832068 Chr.2: 152607397-152607397 on Build GRCh38
rs769076905 Chr.2: 152607332-152607332 on Build GRCh38
rs773309813 Chr.2: 152607372-152607372 on Build GRCh38
rs976353693 Chr.2: 152607505-152607508 on Build GRCh38
rs753611923 Chr.2: 152607400-152607400 on Build GRCh38
rs397839032 Chr.2: 152607505-152607506 on Build GRCh38
rs1866115 Chr.2: 152607494-152607494 on Build GRCh38
rs931422280 Chr.2: 152607496-152607496 on Build GRCh38
rs778481687 Chr.2: 152607411-152607411 on Build GRCh38
rs573607400 Chr.2: 152607508-152607508 on Build GRCh38
rs377497365 Chr.2: 152607358-152607358 on Build GRCh38
rs934086543 Chr.2: 152607507-152607508 on Build GRCh38
rs781579764 Chr.2: 152607446-152607446 on Build GRCh38
rs747462149 Chr.2: 152607440-152607440 on Build GRCh38

View Full Product Details