Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 134478533-134478806 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANAPC13
Entrez Gene ID
25847
Gene Name
anaphase promoting complex subunit 13
Gene Aliases
APC13, SWM1
UniGene
Hs.106909
Gene Chromosome Location
Chr.3: 134477704-134486023 on Build GRCh38
Transcripts
NM_001242374.1 NM_001242375.1 NM_015391.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
22078 c.171G>A p.W57* Chr.3: 134478644-134478644 on Build GRCh38
728318 c.165G>A p.M55I Chr.3: 134478650-134478650 on Build GRCh38
7264716 c.176A>C p.D59A Chr.3: 134478639-134478639 on Build GRCh38
5863576 c.146C>T p.S49F Chr.3: 134478669-134478669 on Build GRCh38
6599527 c.136A>G p.T46A Chr.3: 134478679-134478679 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1053814668 Chr.3: 134478761-134478761 on Build GRCh38
rs576647857 Chr.3: 134478706-134478706 on Build GRCh38
rs1019039483 Chr.3: 134478710-134478710 on Build GRCh38
rs780666302 Chr.3: 134478619-134478619 on Build GRCh38
rs114533137 Chr.3: 134478589-134478589 on Build GRCh38
rs779259723 Chr.3: 134478726-134478726 on Build GRCh38
rs780316486 Chr.3: 134478730-134478730 on Build GRCh38
rs573927795 Chr.3: 134478603-134478603 on Build GRCh38
rs150028607 Chr.3: 134478568-134478568 on Build GRCh38
rs546745783 Chr.3: 134478653-134478653 on Build GRCh38
rs754808129 Chr.3: 134478571-134478571 on Build GRCh38
rs1000849571 Chr.3: 134478736-134478736 on Build GRCh38
rs202107313 Chr.3: 134478654-134478654 on Build GRCh38
rs11544840 Chr.3: 134478669-134478669 on Build GRCh38
rs758839880 Chr.3: 134478729-134478729 on Build GRCh38
rs376148183 Chr.3: 134478694-134478694 on Build GRCh38
rs753976604 Chr.3: 134478716-134478716 on Build GRCh38
rs56203660 Chr.3: 134478724-134478724 on Build GRCh38
rs368766554 Chr.3: 134478583-134478583 on Build GRCh38
rs11544841 Chr.3: 134478634-134478634 on Build GRCh38
rs895160267 Chr.3: 134478771-134478771 on Build GRCh38
rs923410147 Chr.3: 134478663-134478663 on Build GRCh38
rs61748112 Chr.3: 134478587-134478587 on Build GRCh38
rs772450733 Chr.3: 134478612-134478612 on Build GRCh38
rs201280783 Chr.3: 134478684-134478684 on Build GRCh38
rs750736411 Chr.3: 134478728-134478728 on Build GRCh38
rs545226312 Chr.3: 134478732-134478732 on Build GRCh38
rs371589483 Chr.3: 134478683-134478683 on Build GRCh38
rs369845547 Chr.3: 134478639-134478639 on Build GRCh38
rs764219659 Chr.3: 134478713-134478713 on Build GRCh38
rs368797631 Chr.3: 134478748-134478748 on Build GRCh38
rs151170068 Chr.3: 134478588-134478588 on Build GRCh38
rs748551766 Chr.3: 134478565-134478565 on Build GRCh38
rs908949263 Chr.3: 134478609-134478609 on Build GRCh38
rs776152451 Chr.3: 134478600-134478616 on Build GRCh38
rs777580159 Chr.3: 134478620-134478620 on Build GRCh38
rs116673007 Chr.3: 134478564-134478564 on Build GRCh38
rs1008456402 Chr.3: 134478686-134478686 on Build GRCh38
rs752720001 Chr.3: 134478701-134478701 on Build GRCh38
rs767558674 Chr.3: 134478574-134478574 on Build GRCh38
rs752708020 Chr.3: 134478575-134478575 on Build GRCh38
rs757351471 Chr.3: 134478721-134478721 on Build GRCh38
rs1043024259 Chr.3: 134478727-134478727 on Build GRCh38
rs145332961 Chr.3: 134478656-134478656 on Build GRCh38
rs200221769 Chr.3: 134478637-134478637 on Build GRCh38

View Full Product Details