Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 64602643-64602878 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CACHD1
Entrez Gene ID
57685
Gene Name
cache domain containing 1
Gene Aliases
-
UniGene
Hs.443891
Gene Chromosome Location
Chr.1: 64470793-64693058 on Build GRCh38
Transcripts
NM_001293274.1 NM_020925.2 XM_011541862.1 XM_011541863.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7315807 c.326A>G p.N109S Chr.1: 64602874-64602874 on Build GRCh38
6617296 c.460C>T p.R154* Chr.1: 64602855-64602855 on Build GRCh38
4668304 c.308G>A p.R103Q Chr.1: 64602856-64602856 on Build GRCh38
7315808 c.479A>G p.N160S Chr.1: 64602874-64602874 on Build GRCh38
6617287 c.414C>T p.F138F Chr.1: 64602809-64602809 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773192353 Chr.1: 64602780-64602780 on Build GRCh38
rs559035954 Chr.1: 64602709-64602709 on Build GRCh38
rs762999785 Chr.1: 64602818-64602819 on Build GRCh38
rs761632257 Chr.1: 64602754-64602754 on Build GRCh38
rs1041333180 Chr.1: 64602797-64602797 on Build GRCh38
rs934834207 Chr.1: 64602842-64602842 on Build GRCh38
rs756889525 Chr.1: 64602789-64602789 on Build GRCh38
rs929783401 Chr.1: 64602822-64602822 on Build GRCh38
rs932018323 Chr.1: 64602730-64602730 on Build GRCh38
rs780741848 Chr.1: 64602791-64602791 on Build GRCh38
rs1055140432 Chr.1: 64602846-64602846 on Build GRCh38
rs749876404 Chr.1: 64602829-64602829 on Build GRCh38
rs757886843 Chr.1: 64602764-64602764 on Build GRCh38
rs952181064 Chr.1: 64602720-64602720 on Build GRCh38
rs867681376 Chr.1: 64602771-64602771 on Build GRCh38
rs943988425 Chr.1: 64602796-64602796 on Build GRCh38
rs527836063 Chr.1: 64602809-64602809 on Build GRCh38
rs774620530 Chr.1: 64602835-64602835 on Build GRCh38
rs780694131 Chr.1: 64602826-64602826 on Build GRCh38
rs768825442 Chr.1: 64602830-64602830 on Build GRCh38
rs1026884179 Chr.1: 64602710-64602710 on Build GRCh38
rs372693053 Chr.1: 64602783-64602783 on Build GRCh38
rs746179035 Chr.1: 64602802-64602802 on Build GRCh38
rs796399258 Chr.1: 64602785-64602785 on Build GRCh38
rs897016479 Chr.1: 64602815-64602815 on Build GRCh38
rs777327603 Chr.1: 64602768-64602768 on Build GRCh38
rs961297219 Chr.1: 64602701-64602701 on Build GRCh38
rs752641787 Chr.1: 64602759-64602759 on Build GRCh38
rs957078708 Chr.1: 64602699-64602699 on Build GRCh38
rs776598960 Chr.1: 64602750-64602750 on Build GRCh38
rs991496993 Chr.1: 64602732-64602732 on Build GRCh38

View Full Product Details