Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 37309209-37309436 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYTH4
Entrez Gene ID
27128
Gene Name
cytohesin 4
Gene Aliases
CYT4, DJ63G5.1, PSCD4
UniGene
Hs.170944
Gene Chromosome Location
Chr.22: 37282383-37315348 on Build GRCh38
Transcripts
NM_001318024.1 NM_013385.4 XM_011530147.2 XM_011530148.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6290390 c.771C>T p.F257F Chr.22: 37309286-37309286 on Build GRCh38
5859317 c.781G>A p.D261N Chr.22: 37309296-37309296 on Build GRCh38
3150306 c.744C>T p.D248D Chr.22: 37309259-37309259 on Build GRCh38
5930097 c.761C>A p.T254N Chr.22: 37309276-37309276 on Build GRCh38
4506905 c.732C>T p.S244S Chr.22: 37309247-37309247 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs542894219 Chr.22: 37309284-37309284 on Build GRCh38
rs750801433 Chr.22: 37309371-37309371 on Build GRCh38
rs535579793 Chr.22: 37309272-37309272 on Build GRCh38
rs747981080 Chr.22: 37309347-37309347 on Build GRCh38
rs905710195 Chr.22: 37309383-37309383 on Build GRCh38
rs565205064 Chr.22: 37309398-37309398 on Build GRCh38
rs375591759 Chr.22: 37309354-37309354 on Build GRCh38
rs780456860 Chr.22: 37309373-37309373 on Build GRCh38
rs757653238 Chr.22: 37309282-37309282 on Build GRCh38
rs866864287 Chr.22: 37309390-37309390 on Build GRCh38
rs369653989 Chr.22: 37309356-37309356 on Build GRCh38
rs774401543 Chr.22: 37309242-37309242 on Build GRCh38
rs199573505 Chr.22: 37309368-37309368 on Build GRCh38
rs778447993 Chr.22: 37309344-37309346 on Build GRCh38
rs764613251 Chr.22: 37309278-37309278 on Build GRCh38
rs749364131 Chr.22: 37309232-37309232 on Build GRCh38
rs1001643471 Chr.22: 37309259-37309259 on Build GRCh38
rs774452887 Chr.22: 37309345-37309345 on Build GRCh38
rs768764540 Chr.22: 37309236-37309236 on Build GRCh38
rs752011000 Chr.22: 37309280-37309280 on Build GRCh38
rs760936304 Chr.22: 37309353-37309353 on Build GRCh38
rs756471561 Chr.22: 37309372-37309372 on Build GRCh38
rs760182971 Chr.22: 37309274-37309274 on Build GRCh38
rs146483512 Chr.22: 37309384-37309384 on Build GRCh38
rs145053771 Chr.22: 37309277-37309277 on Build GRCh38
rs764666438 Chr.22: 37309366-37309366 on Build GRCh38
rs55891452 Chr.22: 37309296-37309296 on Build GRCh38
rs749291843 Chr.22: 37309413-37309413 on Build GRCh38
rs749421525 Chr.22: 37309334-37309334 on Build GRCh38
rs767168826 Chr.22: 37309271-37309271 on Build GRCh38
rs780403616 Chr.22: 37309330-37309330 on Build GRCh38
rs757771054 Chr.22: 37309369-37309369 on Build GRCh38
rs376617044 Chr.22: 37309337-37309337 on Build GRCh38
rs755951276 Chr.22: 37309362-37309362 on Build GRCh38
rs766017763 Chr.22: 37309275-37309275 on Build GRCh38
rs973665866 Chr.22: 37309326-37309333 on Build GRCh38
rs561519220 Chr.22: 37309326-37309326 on Build GRCh38
rs376928228 Chr.22: 37309350-37309350 on Build GRCh38
rs371595811 Chr.22: 37309348-37309348 on Build GRCh38
rs761700668 Chr.22: 37309260-37309260 on Build GRCh38
rs777531673 Chr.22: 37309405-37309405 on Build GRCh38
rs369126022 Chr.22: 37309351-37309351 on Build GRCh38
rs746238224 Chr.22: 37309300-37309300 on Build GRCh38
rs768053765 Chr.22: 37309370-37309370 on Build GRCh38
rs768817291 Chr.22: 37309344-37309344 on Build GRCh38
rs964007352 Chr.22: 37309314-37309314 on Build GRCh38

View Full Product Details