Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 59970017-59970288 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CDH26
Entrez Gene ID
60437
Gene Name
cadherin 26
Gene Aliases
VR20
UniGene
Hs.729046
Gene Chromosome Location
Chr.20: 59957764-60035629 on Build GRCh38
Transcripts
NM_177980.2 XM_011528969.2 XM_017027994.1 XM_017027995.1 XM_017027996.1 XM_017027997.1 XM_017027998.1 XM_017027999.1 XM_017028000.1 XM_017028001.1 XM_017028002.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6625046 c.152G>A p.R51Q Chr.20: 59970107-59970107 on Build GRCh38
1307564 c.156C>T p.S52S Chr.20: 59970111-59970111 on Build GRCh38
1307562 c.149G>A p.R50Q Chr.20: 59970104-59970104 on Build GRCh38
4099914 c.184G>A p.E62K Chr.20: 59970139-59970139 on Build GRCh38
6278262 c.148C>T p.R50W Chr.20: 59970103-59970103 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs758545172 Chr.20: 59970106-59970106 on Build GRCh38
rs770521594 Chr.20: 59970082-59970082 on Build GRCh38
rs752186363 Chr.20: 59970090-59970090 on Build GRCh38
rs769655102 Chr.20: 59970063-59970063 on Build GRCh38
rs777169775 Chr.20: 59970164-59970164 on Build GRCh38
rs1045814832 Chr.20: 59970216-59970216 on Build GRCh38
rs367835493 Chr.20: 59970194-59970194 on Build GRCh38
rs892999704 Chr.20: 59970241-59970241 on Build GRCh38
rs750689246 Chr.20: 59970189-59970189 on Build GRCh38
rs760219581 Chr.20: 59970155-59970155 on Build GRCh38
rs775218677 Chr.20: 59970231-59970231 on Build GRCh38
rs759025673 Chr.20: 59970088-59970088 on Build GRCh38
rs929701036 Chr.20: 59970213-59970213 on Build GRCh38
rs148524136 Chr.20: 59970147-59970147 on Build GRCh38
rs985702964 Chr.20: 59970119-59970119 on Build GRCh38
rs144755899 Chr.20: 59970142-59970142 on Build GRCh38
rs761276555 Chr.20: 59970092-59970092 on Build GRCh38
rs770745070 Chr.20: 59970131-59970131 on Build GRCh38
rs774025399 Chr.20: 59970084-59970084 on Build GRCh38
rs1045441689 Chr.20: 59970243-59970243 on Build GRCh38
rs371260056 Chr.20: 59970127-59970127 on Build GRCh38
rs1048781727 Chr.20: 59970236-59970236 on Build GRCh38
rs536661438 Chr.20: 59970103-59970103 on Build GRCh38
rs371501209 Chr.20: 59970238-59970238 on Build GRCh38
rs374301665 Chr.20: 59970191-59970191 on Build GRCh38
rs151039129 Chr.20: 59970132-59970132 on Build GRCh38
rs906837448 Chr.20: 59970218-59970218 on Build GRCh38
rs762146610 Chr.20: 59970175-59970175 on Build GRCh38
rs921733449 Chr.20: 59970134-59970134 on Build GRCh38
rs745537184 Chr.20: 59970138-59970138 on Build GRCh38
rs530921974 Chr.20: 59970177-59970177 on Build GRCh38
rs940343867 Chr.20: 59970239-59970239 on Build GRCh38
rs764644570 Chr.20: 59970094-59970094 on Build GRCh38
rs373877280 Chr.20: 59970071-59970071 on Build GRCh38
rs765192392 Chr.20: 59970144-59970144 on Build GRCh38
rs765633969 Chr.20: 59970188-59970188 on Build GRCh38
rs368082728 Chr.20: 59970122-59970122 on Build GRCh38
rs754182983 Chr.20: 59970102-59970102 on Build GRCh38
rs577785384 Chr.20: 59970192-59970192 on Build GRCh38
rs777728621 Chr.20: 59970124-59970124 on Build GRCh38
rs777621961 Chr.20: 59970232-59970232 on Build GRCh38
rs747214170 Chr.20: 59970211-59970212 on Build GRCh38
rs771120841 Chr.20: 59970212-59970213 on Build GRCh38
rs776231597 Chr.20: 59970104-59970104 on Build GRCh38
rs762762865 Chr.20: 59970075-59970075 on Build GRCh38
rs764734372 Chr.20: 59970159-59970159 on Build GRCh38
rs755021514 Chr.20: 59970220-59970220 on Build GRCh38
rs1001873959 Chr.20: 59970224-59970224 on Build GRCh38
rs978494585 Chr.20: 59970079-59970079 on Build GRCh38
rs748076277 Chr.20: 59970052-59970052 on Build GRCh38
rs78153451 Chr.20: 59970262-59970262 on Build GRCh38
rs751670276 Chr.20: 59970212-59970212 on Build GRCh38
rs143651384 Chr.20: 59970089-59970089 on Build GRCh38
rs554586698 Chr.20: 59970107-59970107 on Build GRCh38

View Full Product Details