Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 172038266-172038505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM3
Entrez Gene ID
26052
Gene Name
dynamin 3
Gene Aliases
Dyna III
UniGene
Hs.654775
Gene Chromosome Location
Chr.1: 171841478-172418466 on Build GRCh38
Transcripts
NM_001136127.2 NM_001278252.1 NM_015569.4 XM_005245079.1 XM_005245080.1 XM_006711268.2 XM_017000976.1 XM_017000977.1 XM_017000978.1 XM_017000979.1 XM_017000980.1 XM_017000981.1 XM_017000982.1 XM_017000983.1 XM_017000984.1 XM_017000985.1 XM_017000986.1 XM_017000987.1 XM_017000988.1 XM_017000989.1 XM_017000990.1 XM_017000991.1 XM_017000992.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3478366 c.947A>T p.N316I Chr.1: 172038416-172038416 on Build GRCh38
4617207 c.913C>T p.L305F Chr.1: 172038382-172038382 on Build GRCh38
365983 c.891G>T p.R297S Chr.1: 172038360-172038360 on Build GRCh38
3478365 c.891G>A p.R297R Chr.1: 172038360-172038360 on Build GRCh38
4402510 c.970A>G p.R324G Chr.1: 172038439-172038439 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751536300 Chr.1: 172038422-172038422 on Build GRCh38
rs759286939 Chr.1: 172038391-172038391 on Build GRCh38
rs746990166 Chr.1: 172038324-172038324 on Build GRCh38
rs763267475 Chr.1: 172038370-172038370 on Build GRCh38
rs1043971124 Chr.1: 172038341-172038341 on Build GRCh38
rs377404194 Chr.1: 172038426-172038426 on Build GRCh38
rs752474386 Chr.1: 172038312-172038312 on Build GRCh38
rs780203628 Chr.1: 172038447-172038447 on Build GRCh38
rs563610918 Chr.1: 172038342-172038342 on Build GRCh38
rs369046433 Chr.1: 172038461-172038461 on Build GRCh38
rs753499710 Chr.1: 172038298-172038298 on Build GRCh38
rs993199587 Chr.1: 172038400-172038400 on Build GRCh38
rs892136603 Chr.1: 172038334-172038334 on Build GRCh38
rs774819321 Chr.1: 172038387-172038387 on Build GRCh38
rs762348915 Chr.1: 172038389-172038389 on Build GRCh38
rs758298192 Chr.1: 172038315-172038315 on Build GRCh38
rs765909922 Chr.1: 172038394-172038394 on Build GRCh38
rs775698504 Chr.1: 172038361-172038361 on Build GRCh38
rs552393073 Chr.1: 172038425-172038425 on Build GRCh38
rs777802924 Chr.1: 172038317-172038317 on Build GRCh38
rs769034475 Chr.1: 172038384-172038384 on Build GRCh38
rs199716360 Chr.1: 172038395-172038395 on Build GRCh38
rs904121277 Chr.1: 172038357-172038357 on Build GRCh38
rs756253027 Chr.1: 172038438-172038438 on Build GRCh38
rs754655032 Chr.1: 172038299-172038299 on Build GRCh38
rs758244650 Chr.1: 172038413-172038413 on Build GRCh38
rs548635608 Chr.1: 172038337-172038337 on Build GRCh38
rs769846099 Chr.1: 172038360-172038360 on Build GRCh38
rs764019043 Chr.1: 172038416-172038416 on Build GRCh38
rs373211480 Chr.1: 172038311-172038311 on Build GRCh38
rs781193654 Chr.1: 172038338-172038338 on Build GRCh38
rs752419373 Chr.1: 172038408-172038408 on Build GRCh38
rs764755733 Chr.1: 172038407-172038407 on Build GRCh38
rs757382462 Chr.1: 172038406-172038406 on Build GRCh38

View Full Product Details