Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 65920255-65920484 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTNNA3
Entrez Gene ID
29119
Gene Name
catenin alpha 3
Gene Aliases
ARVD13, VR22
UniGene
Hs.21375
Gene Chromosome Location
Chr.10: 65912518-67696217 on Build GRCh38
Transcripts
NM_001127384.2 NM_013266.3 XM_017016151.1 XM_017016152.1 XM_017016153.1 XM_017016154.1 XM_017016155.1 XM_017016156.1 XM_017016157.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6637561 c.2586G>T p.E862D Chr.10: 65920432-65920432 on Build GRCh38
684710 c.2653G>T p.V885F Chr.10: 65920365-65920365 on Build GRCh38
3709929 c.2658G>T p.M886I Chr.10: 65920360-65920360 on Build GRCh38
684706 c.2574G>T p.L858F Chr.10: 65920444-65920444 on Build GRCh38
1249394 c.2543G>A p.W848* Chr.10: 65920475-65920475 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766896063 Chr.10: 65920443-65920443 on Build GRCh38
rs766935847 Chr.10: 65920408-65920408 on Build GRCh38
rs587782935 Chr.10: 65920447-65920447 on Build GRCh38
rs767799982 Chr.10: 65920450-65920450 on Build GRCh38
rs867483256 Chr.10: 65920356-65920356 on Build GRCh38
rs762187965 Chr.10: 65920328-65920328 on Build GRCh38
rs755208593 Chr.10: 65920449-65920449 on Build GRCh38
rs369681914 Chr.10: 65920315-65920315 on Build GRCh38
rs536070806 Chr.10: 65920301-65920301 on Build GRCh38
rs374368635 Chr.10: 65920389-65920389 on Build GRCh38
rs766707232 Chr.10: 65920369-65920369 on Build GRCh38
rs1002623045 Chr.10: 65920351-65920351 on Build GRCh38
rs751884087 Chr.10: 65920371-65920371 on Build GRCh38
rs761449098 Chr.10: 65920401-65920401 on Build GRCh38
rs377740786 Chr.10: 65920390-65920390 on Build GRCh38
rs768330893 Chr.10: 65920415-65920415 on Build GRCh38
rs771653307 Chr.10: 65920288-65920288 on Build GRCh38
rs730880073 Chr.10: 65920430-65920430 on Build GRCh38
rs773840516 Chr.10: 65920432-65920432 on Build GRCh38
rs761152565 Chr.10: 65920379-65920380 on Build GRCh38
rs761402440 Chr.10: 65920418-65920418 on Build GRCh38
rs376134614 Chr.10: 65920362-65920362 on Build GRCh38
rs190581449 Chr.10: 65920322-65920322 on Build GRCh38
rs750697572 Chr.10: 65920341-65920341 on Build GRCh38
rs544334804 Chr.10: 65920437-65920437 on Build GRCh38
rs557828885 Chr.10: 65920461-65920461 on Build GRCh38
rs765248471 Chr.10: 65920460-65920468 on Build GRCh38
rs1026178943 Chr.10: 65920438-65920438 on Build GRCh38
rs773632703 Chr.10: 65920304-65920304 on Build GRCh38
rs41313840 Chr.10: 65920445-65920445 on Build GRCh38
rs763644062 Chr.10: 65920321-65920321 on Build GRCh38
rs759618368 Chr.10: 65920449-65920450 on Build GRCh38
rs906568114 Chr.10: 65920331-65920331 on Build GRCh38
rs778921417 Chr.10: 65920400-65920400 on Build GRCh38
rs765799315 Chr.10: 65920338-65920338 on Build GRCh38
rs758842998 Chr.10: 65920364-65920364 on Build GRCh38
rs775103016 Chr.10: 65920402-65920402 on Build GRCh38
rs575703238 Chr.10: 65920417-65920417 on Build GRCh38
rs771473515 Chr.10: 65920388-65920388 on Build GRCh38
rs781167292 Chr.10: 65920388-65920388 on Build GRCh38
rs746704810 Chr.10: 65920404-65920404 on Build GRCh38
rs755081926 Chr.10: 65920373-65920373 on Build GRCh38
rs752661010 Chr.10: 65920456-65920456 on Build GRCh38

View Full Product Details