Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 62813837-62814110 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EGR2
Entrez Gene ID
1959
Gene Name
early growth response 2
Gene Aliases
AT591, CMT1D, CMT4E, KROX20
UniGene
Hs.1395
Gene Chromosome Location
Chr.10: 62811996-62819167 on Build GRCh38
Transcripts
NM_000399.4 NM_001136177.2 NM_001136178.1 NM_001136179.2 NM_001321037.1 XM_011539427.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4599061 c.772G>C p.V258L Chr.10: 62813866-62813866 on Build GRCh38
6652346 c.731C>A p.P244Q Chr.10: 62813907-62813907 on Build GRCh38
1348562 c.559G>A p.A187T Chr.10: 62814079-62814079 on Build GRCh38
1505934 c.616C>T p.P206S Chr.10: 62814022-62814022 on Build GRCh38
684741 c.754C>A p.P252T Chr.10: 62813884-62813884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368282744 Chr.10: 62813869-62813869 on Build GRCh38
rs758709729 Chr.10: 62813864-62813864 on Build GRCh38
rs755310380 Chr.10: 62813974-62813974 on Build GRCh38
rs749981503 Chr.10: 62813957-62813957 on Build GRCh38
rs1055082588 Chr.10: 62814030-62814030 on Build GRCh38
rs772044885 Chr.10: 62814026-62814026 on Build GRCh38
rs750955966 Chr.10: 62813861-62813861 on Build GRCh38
rs936268533 Chr.10: 62814025-62814025 on Build GRCh38
rs1011150442 Chr.10: 62813927-62813927 on Build GRCh38
rs750792592 Chr.10: 62814041-62814042 on Build GRCh38
rs749421476 Chr.10: 62814013-62814013 on Build GRCh38
rs761429712 Chr.10: 62814089-62814089 on Build GRCh38
rs776165437 Chr.10: 62813945-62813945 on Build GRCh38
rs764745834 Chr.10: 62813956-62813956 on Build GRCh38
rs747371531 Chr.10: 62813872-62813872 on Build GRCh38
rs761250551 Chr.10: 62813948-62813948 on Build GRCh38
rs781325226 Chr.10: 62813885-62813885 on Build GRCh38
rs372491511 Chr.10: 62814078-62814078 on Build GRCh38
rs775350294 Chr.10: 62814029-62814029 on Build GRCh38
rs746763888 Chr.10: 62814035-62814035 on Build GRCh38
rs748179879 Chr.10: 62813897-62813897 on Build GRCh38
rs794727248 Chr.10: 62813959-62813959 on Build GRCh38
rs780441708 Chr.10: 62813868-62813868 on Build GRCh38
rs781408172 Chr.10: 62813993-62813993 on Build GRCh38
rs745823802 Chr.10: 62813905-62813905 on Build GRCh38
rs200910043 Chr.10: 62813874-62813874 on Build GRCh38
rs760456822 Chr.10: 62813922-62813922 on Build GRCh38
rs224083 Chr.10: 62814011-62814011 on Build GRCh38
rs768344815 Chr.10: 62814042-62814042 on Build GRCh38
rs758869303 Chr.10: 62813968-62813968 on Build GRCh38
rs774391305 Chr.10: 62813902-62813902 on Build GRCh38
rs766543445 Chr.10: 62813943-62813943 on Build GRCh38
rs561006361 Chr.10: 62814076-62814076 on Build GRCh38
rs769998360 Chr.10: 62813898-62813898 on Build GRCh38
rs144739493 Chr.10: 62814077-62814077 on Build GRCh38
rs201264628 Chr.10: 62813890-62813890 on Build GRCh38
rs1022830303 Chr.10: 62814008-62814008 on Build GRCh38
rs776212792 Chr.10: 62814074-62814074 on Build GRCh38
rs530614586 Chr.10: 62813973-62813973 on Build GRCh38
rs1021225769 Chr.10: 62813954-62813954 on Build GRCh38
rs139147487 Chr.10: 62813994-62813994 on Build GRCh38
rs776440321 Chr.10: 62813866-62813866 on Build GRCh38
rs775326892 Chr.10: 62813919-62813919 on Build GRCh38
rs766860417 Chr.10: 62813969-62813969 on Build GRCh38
rs771954217 Chr.10: 62813912-62813912 on Build GRCh38
rs769473870 Chr.10: 62814090-62814090 on Build GRCh38
rs763792577 Chr.10: 62813938-62813938 on Build GRCh38

View Full Product Details