Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 77887028-77887297 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADCK1
Entrez Gene ID
57143
Gene Name
aarF domain containing kinase 1
Gene Aliases
-
UniGene
Hs.413208
Gene Chromosome Location
Chr.14: 77800058-77933954 on Build GRCh38
Transcripts
NM_001142545.1 NM_020421.3 XM_005267885.3 XM_005267886.3 XM_011536987.2 XM_011536988.2 XM_017021474.1 XM_017021475.1 XM_017021476.1 XM_017021477.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7227212 c.424-7C>T p.? Chr.14: 77887084-77887084 on Build GRCh38
6343360 c.505G>T p.D169Y Chr.14: 77887172-77887172 on Build GRCh38
5088831 c.424-3C>T p.? Chr.14: 77887088-77887088 on Build GRCh38
7222594 c.469G>T p.A157S Chr.14: 77887136-77887136 on Build GRCh38
4499226 c.333C>T p.H111H Chr.14: 77887204-77887204 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767728616 Chr.14: 77887211-77887211 on Build GRCh38
rs751331821 Chr.14: 77887200-77887200 on Build GRCh38
rs200211943 Chr.14: 77887215-77887215 on Build GRCh38
rs762801002 Chr.14: 77887146-77887146 on Build GRCh38
rs555961756 Chr.14: 77887120-77887120 on Build GRCh38
rs763974286 Chr.14: 77887195-77887195 on Build GRCh38
rs750459181 Chr.14: 77887170-77887170 on Build GRCh38
rs571984075 Chr.14: 77887171-77887171 on Build GRCh38
rs35636358 Chr.14: 77887135-77887135 on Build GRCh38
rs368684141 Chr.14: 77887259-77887259 on Build GRCh38
rs759051550 Chr.14: 77887189-77887189 on Build GRCh38
rs780580464 Chr.14: 77887176-77887176 on Build GRCh38
rs760236958 Chr.14: 77887125-77887125 on Build GRCh38
rs141575665 Chr.14: 77887086-77887086 on Build GRCh38
rs770268465 Chr.14: 77887115-77887115 on Build GRCh38
rs751814939 Chr.14: 77887148-77887148 on Build GRCh38
rs562832907 Chr.14: 77887228-77887228 on Build GRCh38
rs752103173 Chr.14: 77887169-77887170 on Build GRCh38
rs142511548 Chr.14: 77887182-77887182 on Build GRCh38
rs771751822 Chr.14: 77887122-77887122 on Build GRCh38
rs753707142 Chr.14: 77887068-77887068 on Build GRCh38
rs767209213 Chr.14: 77887068-77887068 on Build GRCh38
rs772829815 Chr.14: 77887123-77887123 on Build GRCh38
rs770425160 Chr.14: 77887126-77887126 on Build GRCh38
rs202239054 Chr.14: 77887244-77887244 on Build GRCh38
rs201695094 Chr.14: 77887093-77887093 on Build GRCh38
rs781135913 Chr.14: 77887158-77887158 on Build GRCh38
rs1016706840 Chr.14: 77887130-77887130 on Build GRCh38
rs775354054 Chr.14: 77887242-77887242 on Build GRCh38
rs779266129 Chr.14: 77887227-77887227 on Build GRCh38
rs752470821 Chr.14: 77887141-77887141 on Build GRCh38
rs768655709 Chr.14: 77887254-77887254 on Build GRCh38
rs760267831 Chr.14: 77887207-77887207 on Build GRCh38
rs763220154 Chr.14: 77887114-77887114 on Build GRCh38
rs758720601 Chr.14: 77887083-77887083 on Build GRCh38
rs764456452 Chr.14: 77887147-77887147 on Build GRCh38
rs1029802855 Chr.14: 77887085-77887085 on Build GRCh38
rs780870923 Chr.14: 77887229-77887229 on Build GRCh38
rs762897245 Chr.14: 77887192-77887192 on Build GRCh38
rs768996018 Chr.14: 77887179-77887179 on Build GRCh38
rs144436820 Chr.14: 77887134-77887134 on Build GRCh38
rs754872549 Chr.14: 77887071-77887071 on Build GRCh38
rs915916186 Chr.14: 77887167-77887167 on Build GRCh38
rs368200753 Chr.14: 77887183-77887183 on Build GRCh38
rs757475947 Chr.14: 77887150-77887150 on Build GRCh38
rs531853809 Chr.14: 77887232-77887232 on Build GRCh38
rs777470419 Chr.14: 77887180-77887180 on Build GRCh38
rs371192783 Chr.14: 77887261-77887261 on Build GRCh38
rs368376408 Chr.14: 77887264-77887264 on Build GRCh38
rs766854138 Chr.14: 77887214-77887214 on Build GRCh38
rs764947817 Chr.14: 77887078-77887078 on Build GRCh38
rs540055984 Chr.14: 77887102-77887102 on Build GRCh38
rs760146820 Chr.14: 77887047-77887047 on Build GRCh38
rs139978828 Chr.14: 77887240-77887240 on Build GRCh38
rs766355252 Chr.14: 77887052-77887052 on Build GRCh38
rs56314853 Chr.14: 77887190-77887190 on Build GRCh38
rs769331279 Chr.14: 77887238-77887238 on Build GRCh38
rs540201983 Chr.14: 77887203-77887203 on Build GRCh38
rs757387734 Chr.14: 77887095-77887095 on Build GRCh38
rs532960788 Chr.14: 77887212-77887212 on Build GRCh38
rs752483597 Chr.14: 77887080-77887080 on Build GRCh38
rs200983054 Chr.14: 77887178-77887178 on Build GRCh38

View Full Product Details