Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 120952676-120952913 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C5
Entrez Gene ID
727
Gene Name
complement component 5
Gene Aliases
C5D, C5a, C5b, CPAMD4, ECLZB
UniGene
Hs.494997
Gene Chromosome Location
Chr.9: 120952335-121075174 on Build GRCh38
Transcripts
NM_001317163.1 NM_001735.2 XM_011518980.2 XM_017015102.1 XM_017015103.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3788156 c.4969T>C p.C1657R Chr.9: 120952801-120952801 on Build GRCh38
5989958 c.5005G>A p.E1669K Chr.9: 120952765-120952765 on Build GRCh38
5655621 c.4972C>T p.Q1658* Chr.9: 120952798-120952798 on Build GRCh38
223364 c.4913C>T p.P1638L Chr.9: 120952857-120952857 on Build GRCh38
274290 c.5003C>T p.A1668V Chr.9: 120952767-120952767 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs769270167 Chr.9: 120952835-120952835 on Build GRCh38
rs903206035 Chr.9: 120952815-120952815 on Build GRCh38
rs1015683149 Chr.9: 120952807-120952807 on Build GRCh38
rs748810050 Chr.9: 120952752-120952752 on Build GRCh38
rs770445612 Chr.9: 120952811-120952811 on Build GRCh38
rs755663613 Chr.9: 120952803-120952803 on Build GRCh38
rs41314049 Chr.9: 120952889-120952889 on Build GRCh38
rs963102358 Chr.9: 120952873-120952873 on Build GRCh38
rs140229894 Chr.9: 120952739-120952739 on Build GRCh38
rs747726925 Chr.9: 120952743-120952743 on Build GRCh38
rs750465028 Chr.9: 120952774-120952774 on Build GRCh38
rs778440054 Chr.9: 120952812-120952812 on Build GRCh38
rs765573116 Chr.9: 120952767-120952767 on Build GRCh38
rs761048515 Chr.9: 120952762-120952762 on Build GRCh38
rs762245967 Chr.9: 120952846-120952846 on Build GRCh38
rs763160721 Chr.9: 120952776-120952776 on Build GRCh38
rs780148284 Chr.9: 120952706-120952706 on Build GRCh38
rs758052311 Chr.9: 120952765-120952765 on Build GRCh38
rs769554427 Chr.9: 120952745-120952745 on Build GRCh38
rs750591495 Chr.9: 120952702-120952702 on Build GRCh38
rs143531056 Chr.9: 120952753-120952753 on Build GRCh38
rs766537320 Chr.9: 120952881-120952881 on Build GRCh38
rs371917232 Chr.9: 120952858-120952858 on Build GRCh38
rs145613662 Chr.9: 120952821-120952821 on Build GRCh38
rs867521048 Chr.9: 120952806-120952806 on Build GRCh38
rs777405924 Chr.9: 120952808-120952808 on Build GRCh38
rs746765096 Chr.9: 120952714-120952714 on Build GRCh38
rs1030615042 Chr.9: 120952829-120952829 on Build GRCh38
rs780846073 Chr.9: 120952800-120952800 on Build GRCh38
rs754753053 Chr.9: 120952722-120952722 on Build GRCh38
rs535056477 Chr.9: 120952763-120952763 on Build GRCh38
rs780210834 Chr.9: 120952817-120952817 on Build GRCh38
rs754984240 Chr.9: 120952795-120952795 on Build GRCh38
rs374914766 Chr.9: 120952759-120952759 on Build GRCh38
rs772694883 Chr.9: 120952748-120952748 on Build GRCh38
rs751429885 Chr.9: 120952787-120952787 on Build GRCh38
rs747007843 Chr.9: 120952826-120952826 on Build GRCh38
rs776806187 Chr.9: 120952839-120952839 on Build GRCh38
rs779625570 Chr.9: 120952802-120952802 on Build GRCh38
rs1054886026 Chr.9: 120952721-120952721 on Build GRCh38
rs953847247 Chr.9: 120952863-120952863 on Build GRCh38
rs766370744 Chr.9: 120952786-120952786 on Build GRCh38
rs765374255 Chr.9: 120952700-120952700 on Build GRCh38
rs201100938 Chr.9: 120952703-120952703 on Build GRCh38
rs763315652 Chr.9: 120952866-120952866 on Build GRCh38
rs748755309 Chr.9: 120952810-120952810 on Build GRCh38
rs369355126 Chr.9: 120952853-120952853 on Build GRCh38

View Full Product Details