Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 299336-299567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFITM5
Entrez Gene ID
387733
Gene Name
interferon induced transmembrane protein 5
Gene Aliases
BRIL, DSPA1, Hrmp1, OI5, fragilis4
UniGene
Hs.443469
Gene Chromosome Location
Chr.11: 298200-299526 on Build GRCh38
Transcripts
NM_001025295.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4987664 c.41C>T p.T14M Chr.11: 299450-299450 on Build GRCh38
5623321 c.121G>A p.V41M Chr.11: 299370-299370 on Build GRCh38
5717547 c.25G>A p.D9N Chr.11: 299466-299466 on Build GRCh38
4032285 c.148T>C p.C50R Chr.11: 299343-299343 on Build GRCh38
6312370 c.133C>A p.L45I Chr.11: 299358-299358 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs138776304 Chr.11: 299390-299390 on Build GRCh38
rs537904760 Chr.11: 299469-299469 on Build GRCh38
rs12803955 Chr.11: 299506-299506 on Build GRCh38
rs57285449 Chr.11: 299411-299411 on Build GRCh38
rs766631084 Chr.11: 299360-299360 on Build GRCh38
rs768759395 Chr.11: 299383-299385 on Build GRCh38
rs762079301 Chr.11: 299459-299459 on Build GRCh38
rs587776916 Chr.11: 299504-299504 on Build GRCh38
rs755699416 Chr.11: 299470-299470 on Build GRCh38
rs373183215 Chr.11: 299541-299541 on Build GRCh38
rs779959262 Chr.11: 299401-299401 on Build GRCh38
rs79625057 Chr.11: 299371-299371 on Build GRCh38
rs765481203 Chr.11: 299408-299408 on Build GRCh38
rs753297728 Chr.11: 299472-299472 on Build GRCh38
rs372443030 Chr.11: 299404-299404 on Build GRCh38
rs866030579 Chr.11: 299488-299488 on Build GRCh38
rs376963969 Chr.11: 299483-299483 on Build GRCh38
rs753298359 Chr.11: 299373-299373 on Build GRCh38
rs573436645 Chr.11: 299501-299501 on Build GRCh38
rs786201032 Chr.11: 299372-299372 on Build GRCh38
rs776714503 Chr.11: 299437-299437 on Build GRCh38
rs529826939 Chr.11: 299546-299546 on Build GRCh38
rs552104680 Chr.11: 299423-299423 on Build GRCh38
rs575370138 Chr.11: 299524-299524 on Build GRCh38
rs369340601 Chr.11: 299395-299395 on Build GRCh38
rs899037993 Chr.11: 299537-299537 on Build GRCh38
rs770142605 Chr.11: 299447-299447 on Build GRCh38
rs146230729 Chr.11: 299400-299400 on Build GRCh38
rs767982316 Chr.11: 299465-299465 on Build GRCh38
rs74557511 Chr.11: 299471-299471 on Build GRCh38
rs767789172 Chr.11: 299421-299429 on Build GRCh38
rs762124752 Chr.11: 299405-299405 on Build GRCh38
rs771942676 Chr.11: 299436-299436 on Build GRCh38
rs142041432 Chr.11: 299396-299396 on Build GRCh38
rs567857926 Chr.11: 299480-299480 on Build GRCh38
rs746160849 Chr.11: 299440-299440 on Build GRCh38
rs568880098 Chr.11: 299457-299457 on Build GRCh38
rs545346603 Chr.11: 299374-299374 on Build GRCh38
rs531009160 Chr.11: 299391-299391 on Build GRCh38
rs113160876 Chr.11: 299517-299517 on Build GRCh38
rs753826000 Chr.11: 299370-299370 on Build GRCh38
rs1014850776 Chr.11: 299533-299533 on Build GRCh38
rs754531975 Chr.11: 299479-299479 on Build GRCh38
rs755605446 Chr.11: 299418-299418 on Build GRCh38
rs986038826 Chr.11: 299538-299538 on Build GRCh38
rs776961228 Chr.11: 299406-299406 on Build GRCh38
rs201497459 Chr.11: 299405-299405 on Build GRCh38
rs747878124 Chr.11: 299433-299433 on Build GRCh38
rs779478631 Chr.11: 299419-299419 on Build GRCh38
rs763485576 Chr.11: 299409-299409 on Build GRCh38
rs770089325 Chr.11: 299500-299500 on Build GRCh38
rs768597079 Chr.11: 299402-299402 on Build GRCh38
rs750152870 Chr.11: 299413-299413 on Build GRCh38
rs200046984 Chr.11: 299450-299450 on Build GRCh38
rs774773661 Chr.11: 299396-299396 on Build GRCh38
rs374800601 Chr.11: 299424-299424 on Build GRCh38
rs544276666 Chr.11: 299540-299540 on Build GRCh38
rs200674387 Chr.11: 299496-299496 on Build GRCh38
rs182560634 Chr.11: 299545-299545 on Build GRCh38
rs373575422 Chr.11: 299454-299454 on Build GRCh38
rs777569246 Chr.11: 299489-299489 on Build GRCh38
rs748186534 Chr.11: 299482-299482 on Build GRCh38
rs545096603 Chr.11: 299503-299503 on Build GRCh38
rs376004295 Chr.11: 299358-299358 on Build GRCh38
rs995087738 Chr.11: 299516-299516 on Build GRCh38
rs1011303178 Chr.11: 299387-299387 on Build GRCh38
rs778848693 Chr.11: 299427-299427 on Build GRCh38
rs915452062 Chr.11: 299460-299460 on Build GRCh38
rs775669597 Chr.11: 299449-299449 on Build GRCh38
rs751820661 Chr.11: 299410-299410 on Build GRCh38

View Full Product Details