Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 67291226-67291497 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCTD19
Entrez Gene ID
146212
Gene Name
potassium channel tetramerization domain containing 19
Gene Aliases
-
UniGene
Hs.299127
Gene Chromosome Location
Chr.16: 67289490-67326758 on Build GRCh38
Transcripts
NM_001100915.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6144965 c.2487C>A p.D829E Chr.16: 67291387-67291387 on Build GRCh38
4931467 c.2455T>C p.Y819H Chr.16: 67291419-67291419 on Build GRCh38
5819820 c.2549T>A p.L850Q Chr.16: 67291325-67291325 on Build GRCh38
4061831 c.2527A>G p.I843V Chr.16: 67291347-67291347 on Build GRCh38
5851217 c.2565+1G>A p.? Chr.16: 67291308-67291308 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778962115 Chr.16: 67291415-67291415 on Build GRCh38
rs756575700 Chr.16: 67291291-67291291 on Build GRCh38
rs774811468 Chr.16: 67291368-67291369 on Build GRCh38
rs570892420 Chr.16: 67291472-67291472 on Build GRCh38
rs760201103 Chr.16: 67291444-67291444 on Build GRCh38
rs376304788 Chr.16: 67291455-67291455 on Build GRCh38
rs201613304 Chr.16: 67291316-67291316 on Build GRCh38
rs781380041 Chr.16: 67291379-67291379 on Build GRCh38
rs188969016 Chr.16: 67291381-67291381 on Build GRCh38
rs372655932 Chr.16: 67291367-67291367 on Build GRCh38
rs772849200 Chr.16: 67291442-67291442 on Build GRCh38
rs753058016 Chr.16: 67291461-67291461 on Build GRCh38
rs765838053 Chr.16: 67291324-67291324 on Build GRCh38
rs1020494933 Chr.16: 67291246-67291246 on Build GRCh38
rs199535652 Chr.16: 67291453-67291453 on Build GRCh38
rs560133232 Chr.16: 67291417-67291417 on Build GRCh38
rs183853423 Chr.16: 67291283-67291283 on Build GRCh38
rs771463991 Chr.16: 67291279-67291279 on Build GRCh38
rs754265134 Chr.16: 67291297-67291297 on Build GRCh38
rs758553082 Chr.16: 67291343-67291343 on Build GRCh38
rs772010225 Chr.16: 67291422-67291422 on Build GRCh38
rs777736543 Chr.16: 67291357-67291357 on Build GRCh38
rs768307814 Chr.16: 67291392-67291392 on Build GRCh38
rs761428939 Chr.16: 67291274-67291274 on Build GRCh38
rs907494791 Chr.16: 67291459-67291459 on Build GRCh38
rs766821775 Chr.16: 67291263-67291267 on Build GRCh38
rs766059021 Chr.16: 67291295-67291295 on Build GRCh38
rs757485368 Chr.16: 67291370-67291370 on Build GRCh38
rs922830484 Chr.16: 67291317-67291317 on Build GRCh38
rs765498856 Chr.16: 67291457-67291457 on Build GRCh38
rs753112636 Chr.16: 67291329-67291329 on Build GRCh38
rs1022477217 Chr.16: 67291256-67291256 on Build GRCh38
rs967137071 Chr.16: 67291249-67291249 on Build GRCh38

View Full Product Details