Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 98835170-98835443 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GALNT12
Entrez Gene ID
79695
Gene Name
polypeptide N-acetylgalactosaminyltransferase 12
Gene Aliases
CRCS1, GalNAc-T12
UniGene
Hs.47099
Gene Chromosome Location
Chr.9: 98807699-98850081 on Build GRCh38
Transcripts
NM_024642.4 XM_006717287.1 XM_011519018.2 XM_011519020.1 XM_017015133.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7276846 c.918-9_918-8delCT p.? Chr.9: 98835240-98835241 on Build GRCh38
3652245 c.1028C>T p.S343F Chr.9: 98835359-98835359 on Build GRCh38
7285649 c.1002T>C p.V334V Chr.9: 98835333-98835333 on Build GRCh38
275253 c.1021G>A p.E341K Chr.9: 98835352-98835352 on Build GRCh38
3652244 c.1010G>A p.G337E Chr.9: 98835341-98835341 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752958820 Chr.9: 98835345-98835345 on Build GRCh38
rs760520936 Chr.9: 98835293-98835293 on Build GRCh38
rs537015652 Chr.9: 98835254-98835254 on Build GRCh38
rs756614355 Chr.9: 98835348-98835348 on Build GRCh38
rs770442686 Chr.9: 98835245-98835245 on Build GRCh38
rs781443955 Chr.9: 98835230-98835230 on Build GRCh38
rs1032830592 Chr.9: 98835212-98835212 on Build GRCh38
rs778251729 Chr.9: 98835351-98835351 on Build GRCh38
rs755176189 Chr.9: 98835221-98835221 on Build GRCh38
rs748459683 Chr.9: 98835237-98835237 on Build GRCh38
rs769786951 Chr.9: 98835329-98835329 on Build GRCh38
rs751775596 Chr.9: 98835219-98835219 on Build GRCh38
rs773891669 Chr.9: 98835256-98835256 on Build GRCh38
rs763868835 Chr.9: 98835300-98835300 on Build GRCh38
rs781422155 Chr.9: 98835342-98835342 on Build GRCh38
rs778289504 Chr.9: 98835366-98835366 on Build GRCh38
rs377266726 Chr.9: 98835391-98835391 on Build GRCh38
rs758537018 Chr.9: 98835215-98835215 on Build GRCh38
rs776358373 Chr.9: 98835395-98835395 on Build GRCh38
rs776007490 Chr.9: 98835339-98835339 on Build GRCh38
rs749861818 Chr.9: 98835359-98835359 on Build GRCh38
rs778081163 Chr.9: 98835247-98835247 on Build GRCh38
rs768484279 Chr.9: 98835392-98835392 on Build GRCh38
rs201140510 Chr.9: 98835267-98835267 on Build GRCh38
rs751621695 Chr.9: 98835336-98835336 on Build GRCh38
rs749808837 Chr.9: 98835253-98835253 on Build GRCh38
rs200412026 Chr.9: 98835204-98835204 on Build GRCh38
rs775275455 Chr.9: 98835277-98835277 on Build GRCh38
rs139783517 Chr.9: 98835373-98835373 on Build GRCh38
rs267602051 Chr.9: 98835316-98835316 on Build GRCh38
rs763961103 Chr.9: 98835198-98835198 on Build GRCh38
rs199514478 Chr.9: 98835369-98835369 on Build GRCh38
rs766388702 Chr.9: 98835325-98835325 on Build GRCh38
rs761786369 Chr.9: 98835311-98835311 on Build GRCh38
rs371535889 Chr.9: 98835217-98835217 on Build GRCh38
rs775189273 Chr.9: 98835378-98835378 on Build GRCh38
rs1017576138 Chr.9: 98835314-98835314 on Build GRCh38
rs1037385337 Chr.9: 98835297-98835297 on Build GRCh38
rs761684895 Chr.9: 98835412-98835412 on Build GRCh38
rs954825101 Chr.9: 98835280-98835280 on Build GRCh38
rs140471094 Chr.9: 98835306-98835306 on Build GRCh38
rs905756683 Chr.9: 98835190-98835190 on Build GRCh38
rs755162936 Chr.9: 98835341-98835341 on Build GRCh38
rs771421585 Chr.9: 98835360-98835360 on Build GRCh38

View Full Product Details