Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 29574747-29574982 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GIT1
Entrez Gene ID
28964
Gene Name
GIT ArfGAP 1
Gene Aliases
-
UniGene
Hs.514051
Gene Chromosome Location
Chr.17: 29573469-29589592 on Build GRCh38
Transcripts
NM_001085454.1 NM_014030.3 XM_011524684.1 XM_011524685.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
705752 c.2192C>A p.P731Q Chr.17: 29574823-29574823 on Build GRCh38
4065184 c.2260G>A p.A754T Chr.17: 29574755-29574755 on Build GRCh38
3819190 c.2119G>A p.A707T Chr.17: 29574869-29574869 on Build GRCh38
1242315 c.2142T>C p.S714S Chr.17: 29574846-29574846 on Build GRCh38
3819189 c.2146G>A p.A716T Chr.17: 29574869-29574869 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374232508 Chr.17: 29574949-29574949 on Build GRCh38
rs773439194 Chr.17: 29574800-29574800 on Build GRCh38
rs772368684 Chr.17: 29574779-29574779 on Build GRCh38
rs753938693 Chr.17: 29574954-29574954 on Build GRCh38
rs753971394 Chr.17: 29574849-29574849 on Build GRCh38
rs771872677 Chr.17: 29574943-29574943 on Build GRCh38
rs761711973 Chr.17: 29574818-29574818 on Build GRCh38
rs11548560 Chr.17: 29574885-29574885 on Build GRCh38
rs751363864 Chr.17: 29574783-29574784 on Build GRCh38
rs773982835 Chr.17: 29574931-29574931 on Build GRCh38
rs775115811 Chr.17: 29574812-29574812 on Build GRCh38
rs761908309 Chr.17: 29574939-29574939 on Build GRCh38
rs769509316 Chr.17: 29574921-29574921 on Build GRCh38
rs753263205 Chr.17: 29574864-29574864 on Build GRCh38
rs758906145 Chr.17: 29574869-29574869 on Build GRCh38
rs982849609 Chr.17: 29574941-29574941 on Build GRCh38
rs903621709 Chr.17: 29574838-29574838 on Build GRCh38
rs201810676 Chr.17: 29574837-29574837 on Build GRCh38
rs759743937 Chr.17: 29574956-29574956 on Build GRCh38
rs767429818 Chr.17: 29574949-29574949 on Build GRCh38
rs766483401 Chr.17: 29574843-29574843 on Build GRCh38
rs138063413 Chr.17: 29574768-29574768 on Build GRCh38
rs754873963 Chr.17: 29574823-29574823 on Build GRCh38
rs769301186 Chr.17: 29574811-29574811 on Build GRCh38
rs375882746 Chr.17: 29574859-29574859 on Build GRCh38
rs778244686 Chr.17: 29574870-29574870 on Build GRCh38
rs755007238 Chr.17: 29574856-29574856 on Build GRCh38
rs748813318 Chr.17: 29574928-29574928 on Build GRCh38
rs745604727 Chr.17: 29574807-29574807 on Build GRCh38
rs768222957 Chr.17: 29574930-29574930 on Build GRCh38
rs767478368 Chr.17: 29574831-29574831 on Build GRCh38
rs764160074 Chr.17: 29574815-29574815 on Build GRCh38
rs779535073 Chr.17: 29574922-29574922 on Build GRCh38
rs760334398 Chr.17: 29574948-29574948 on Build GRCh38
rs762322741 Chr.17: 29574813-29574813 on Build GRCh38
rs142576554 Chr.17: 29574839-29574839 on Build GRCh38
rs774362777 Chr.17: 29574816-29574816 on Build GRCh38
rs550818 Chr.17: 29574957-29574957 on Build GRCh38
rs992357234 Chr.17: 29574788-29574788 on Build GRCh38
rs969674531 Chr.17: 29574826-29574826 on Build GRCh38
rs773104360 Chr.17: 29574947-29574947 on Build GRCh38
rs547494712 Chr.17: 29574771-29574771 on Build GRCh38

View Full Product Details