Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 120167318-120167543 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPR156
Entrez Gene ID
165829
Gene Name
G protein-coupled receptor 156
Gene Aliases
GABABL, PGR28
UniGene
Hs.333358
Gene Chromosome Location
Chr.3: 120165481-120285122 on Build GRCh38
Transcripts
NM_001168271.1 NM_153002.2 XM_005247165.2 XM_011512484.2 XM_011512487.1 XM_011512488.2 XM_017005795.1 XM_017005796.1 XM_017005797.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6480222 c.2157C>G p.D719E Chr.3: 120167320-120167320 on Build GRCh38
4939590 c.2115C>G p.A705A Chr.3: 120167362-120167362 on Build GRCh38
275345 c.1963C>T p.R655* Chr.3: 120167514-120167514 on Build GRCh38
581155 c.2109C>T p.G703G Chr.3: 120167368-120167368 on Build GRCh38
5821628 c.2123C>A p.P708H Chr.3: 120167354-120167354 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs867700471 Chr.3: 120167415-120167415 on Build GRCh38
rs773109242 Chr.3: 120167367-120167367 on Build GRCh38
rs911897667 Chr.3: 120167379-120167379 on Build GRCh38
rs748735036 Chr.3: 120167350-120167350 on Build GRCh38
rs200587090 Chr.3: 120167427-120167427 on Build GRCh38
rs772131075 Chr.3: 120167499-120167499 on Build GRCh38
rs112549051 Chr.3: 120167475-120167475 on Build GRCh38
rs200929093 Chr.3: 120167387-120167387 on Build GRCh38
rs778461783 Chr.3: 120167459-120167459 on Build GRCh38
rs752144877 Chr.3: 120167501-120167503 on Build GRCh38
rs200604393 Chr.3: 120167469-120167469 on Build GRCh38
rs115973935 Chr.3: 120167419-120167419 on Build GRCh38
rs150001619 Chr.3: 120167513-120167513 on Build GRCh38
rs754544859 Chr.3: 120167396-120167398 on Build GRCh38
rs748905811 Chr.3: 120167441-120167441 on Build GRCh38
rs114542754 Chr.3: 120167418-120167418 on Build GRCh38
rs763087402 Chr.3: 120167405-120167405 on Build GRCh38
rs553212940 Chr.3: 120167514-120167514 on Build GRCh38
rs750786372 Chr.3: 120167422-120167422 on Build GRCh38
rs139313892 Chr.3: 120167511-120167511 on Build GRCh38
rs760612038 Chr.3: 120167386-120167386 on Build GRCh38
rs386665142 Chr.3: 120167418-120167419 on Build GRCh38
rs761772174 Chr.3: 120167366-120167366 on Build GRCh38
rs754620284 Chr.3: 120167455-120167455 on Build GRCh38
rs923653303 Chr.3: 120167449-120167449 on Build GRCh38
rs780411492 Chr.3: 120167431-120167431 on Build GRCh38
rs767034751 Chr.3: 120167460-120167464 on Build GRCh38
rs773739887 Chr.3: 120167361-120167361 on Build GRCh38
rs779752844 Chr.3: 120167346-120167346 on Build GRCh38
rs867162378 Chr.3: 120167342-120167342 on Build GRCh38
rs768056878 Chr.3: 120167421-120167421 on Build GRCh38
rs776319315 Chr.3: 120167520-120167520 on Build GRCh38
rs939480246 Chr.3: 120167407-120167407 on Build GRCh38
rs973736400 Chr.3: 120167360-120167360 on Build GRCh38
rs775719509 Chr.3: 120167388-120167388 on Build GRCh38
rs367758171 Chr.3: 120167357-120167357 on Build GRCh38

View Full Product Details