Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 162344032-162344281 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GCA
Entrez Gene ID
25801
Gene Name
grancalcin
Gene Aliases
GCL
UniGene
Hs.377894
Gene Chromosome Location
Chr.2: 162318768-162371605 on Build GRCh38
Transcripts
NM_012198.3 XM_005246446.2 XM_006712398.3 XM_006712400.3 XM_006712401.3 XM_017003764.1 XM_017003765.1 XM_017003766.1 XM_017003769.1

SNP Detail

SNP ID Chromosome Location
rs778016125 Chr.2: 162344198-162344198 on Build GRCh38
rs989225047 Chr.2: 162344250-162344250 on Build GRCh38
rs534733381 Chr.2: 162344205-162344205 on Build GRCh38
rs994204567 Chr.2: 162344193-162344193 on Build GRCh38
rs986114960 Chr.2: 162344066-162344066 on Build GRCh38
rs754048627 Chr.2: 162344242-162344242 on Build GRCh38
rs372896502 Chr.2: 162344225-162344225 on Build GRCh38
rs982192081 Chr.2: 162344153-162344153 on Build GRCh38
rs775456001 Chr.2: 162344138-162344138 on Build GRCh38
rs749047786 Chr.2: 162344230-162344231 on Build GRCh38
rs7567026 Chr.2: 162344168-162344168 on Build GRCh38
rs772671473 Chr.2: 162344246-162344246 on Build GRCh38
rs996572738 Chr.2: 162344087-162344087 on Build GRCh38
rs955131766 Chr.2: 162344115-162344115 on Build GRCh38
rs762863330 Chr.2: 162344234-162344234 on Build GRCh38
rs763539843 Chr.2: 162344222-162344222 on Build GRCh38
rs1048501665 Chr.2: 162344194-162344194 on Build GRCh38
rs537056010 Chr.2: 162344137-162344137 on Build GRCh38
rs749496273 Chr.2: 162344202-162344202 on Build GRCh38
rs762835855 Chr.2: 162344166-162344166 on Build GRCh38
rs569823913 Chr.2: 162344118-162344118 on Build GRCh38
rs377110441 Chr.2: 162344162-162344162 on Build GRCh38
rs927611334 Chr.2: 162344159-162344159 on Build GRCh38
rs897694811 Chr.2: 162344180-162344180 on Build GRCh38
rs375649533 Chr.2: 162344151-162344151 on Build GRCh38
rs756171068 Chr.2: 162344239-162344239 on Build GRCh38
rs369137863 Chr.2: 162344235-162344235 on Build GRCh38
rs888532210 Chr.2: 162344197-162344197 on Build GRCh38
rs916330929 Chr.2: 162344149-162344149 on Build GRCh38
rs777730174 Chr.2: 162344240-162344240 on Build GRCh38
rs548481242 Chr.2: 162344104-162344104 on Build GRCh38
rs776295741 Chr.2: 162344224-162344224 on Build GRCh38
rs376042341 Chr.2: 162344227-162344227 on Build GRCh38
rs914901038 Chr.2: 162344147-162344147 on Build GRCh38
rs935049464 Chr.2: 162344123-162344123 on Build GRCh38
rs371966041 Chr.2: 162344099-162344099 on Build GRCh38
rs372697444 Chr.2: 162344245-162344245 on Build GRCh38
rs764942681 Chr.2: 162344226-162344226 on Build GRCh38
rs927876138 Chr.2: 162344165-162344165 on Build GRCh38
rs1018489890 Chr.2: 162344076-162344076 on Build GRCh38
rs377430052 Chr.2: 162344259-162344259 on Build GRCh38
rs772767492 Chr.2: 162344207-162344207 on Build GRCh38
rs757623211 Chr.2: 162344243-162344243 on Build GRCh38
rs773790858 Chr.2: 162344206-162344206 on Build GRCh38
rs760267924 Chr.2: 162344221-162344221 on Build GRCh38
rs965633677 Chr.2: 162344214-162344214 on Build GRCh38
rs976917683 Chr.2: 162344116-162344116 on Build GRCh38
rs751482886 Chr.2: 162344236-162344236 on Build GRCh38
rs958178713 Chr.2: 162344143-162344143 on Build GRCh38
rs947784581 Chr.2: 162344152-162344152 on Build GRCh38
rs774844613 Chr.2: 162344210-162344210 on Build GRCh38

View Full Product Details