Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 89013132-89013357 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GBP3
Entrez Gene ID
2635
Gene Name
guanylate binding protein 3
Gene Aliases
-
UniGene
Hs.720167
Gene Chromosome Location
Chr.1: 89006677-89022880 on Build GRCh38
Transcripts
NM_001319179.1 NM_001319180.1 NM_001319181.1 NM_018284.2 NR_135007.1 XM_005270743.2 XM_006710574.2 XM_011541233.1 XM_011541234.1 XM_017001013.1 XM_017001014.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3806053 c.708C>T p.F236F Chr.1: 89013345-89013345 on Build GRCh38
912943 c.709G>A p.D237N Chr.1: 89013344-89013344 on Build GRCh38
5770974 c.736G>T p.A246S Chr.1: 89013317-89013317 on Build GRCh38
6881002 c.862G>A p.G288R Chr.1: 89013191-89013191 on Build GRCh38
83914 c.793G>T p.A265S Chr.1: 89013260-89013260 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764904426 Chr.1: 89013224-89013224 on Build GRCh38
rs746087922 Chr.1: 89013162-89013162 on Build GRCh38
rs778797204 Chr.1: 89013258-89013258 on Build GRCh38
rs753305159 Chr.1: 89013306-89013306 on Build GRCh38
rs773250228 Chr.1: 89013272-89013272 on Build GRCh38
rs750930105 Chr.1: 89013298-89013298 on Build GRCh38
rs759696009 Chr.1: 89013178-89013178 on Build GRCh38
rs769900550 Chr.1: 89013279-89013279 on Build GRCh38
rs751925648 Chr.1: 89013205-89013205 on Build GRCh38
rs1055903709 Chr.1: 89013200-89013200 on Build GRCh38
rs113372144 Chr.1: 89013160-89013160 on Build GRCh38
rs757702853 Chr.1: 89013210-89013210 on Build GRCh38
rs779488259 Chr.1: 89013249-89013249 on Build GRCh38
rs755630445 Chr.1: 89013246-89013246 on Build GRCh38
rs370976551 Chr.1: 89013202-89013202 on Build GRCh38
rs375199876 Chr.1: 89013329-89013329 on Build GRCh38
rs146861483 Chr.1: 89013184-89013184 on Build GRCh38
rs760256930 Chr.1: 89013211-89013212 on Build GRCh38
rs776806694 Chr.1: 89013172-89013172 on Build GRCh38
rs775495318 Chr.1: 89013282-89013282 on Build GRCh38
rs754595573 Chr.1: 89013251-89013251 on Build GRCh38
rs750970249 Chr.1: 89013217-89013217 on Build GRCh38
rs201816821 Chr.1: 89013189-89013189 on Build GRCh38
rs12118276 Chr.1: 89013290-89013290 on Build GRCh38
rs368895425 Chr.1: 89013309-89013309 on Build GRCh38
rs953548671 Chr.1: 89013299-89013299 on Build GRCh38
rs746965247 Chr.1: 89013274-89013274 on Build GRCh38
rs17130686 Chr.1: 89013192-89013192 on Build GRCh38
rs895861174 Chr.1: 89013223-89013223 on Build GRCh38
rs892496829 Chr.1: 89013242-89013242 on Build GRCh38
rs747846082 Chr.1: 89013317-89013317 on Build GRCh38
rs370692800 Chr.1: 89013328-89013328 on Build GRCh38
rs113490655 Chr.1: 89013185-89013185 on Build GRCh38
rs762195381 Chr.1: 89013292-89013292 on Build GRCh38
rs746958235 Chr.1: 89013332-89013332 on Build GRCh38
rs769346921 Chr.1: 89013273-89013273 on Build GRCh38
rs374589463 Chr.1: 89013250-89013250 on Build GRCh38
rs747981376 Chr.1: 89013259-89013259 on Build GRCh38
rs774647388 Chr.1: 89013291-89013291 on Build GRCh38
rs899658696 Chr.1: 89013278-89013278 on Build GRCh38
rs772098411 Chr.1: 89013263-89013263 on Build GRCh38
rs369702754 Chr.1: 89013296-89013296 on Build GRCh38
rs4656076 Chr.1: 89013171-89013171 on Build GRCh38
rs761102669 Chr.1: 89013271-89013271 on Build GRCh38
rs765848735 Chr.1: 89013305-89013305 on Build GRCh38

View Full Product Details