Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 68191188-68191461 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FTLP10
Entrez Gene ID
100130017
Gene Name
ferritin light chain pseudogene 10
Gene Aliases
-
UniGene
Hs.582037
Gene Chromosome Location
Chr.4: 68182292-68212470 on Build GRCh38
Transcripts
NR_015446.1 NR_104048.1

Gene Symbol
TMPRSS11BNL
Entrez Gene ID
401136
Gene Name
TMPRSS11B N-terminal like, pseudogene
Gene Aliases
-
UniGene
Hs.652046
Gene Chromosome Location
Chr.4: 68184129-68218080 on Build GRCh38
Transcripts
NR_015446.1 NR_104048.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6224957 c.287C>A p.S96Y Chr.4: 68191272-68191272 on Build GRCh38
1633752 c.262G>A p.V88I Chr.4: 68191297-68191297 on Build GRCh38
5463362 c.242G>T p.R81I Chr.4: 68191407-68191407 on Build GRCh38
5401430 c.302C>T p.S101L Chr.4: 68191257-68191257 on Build GRCh38
1430629 c.303A>G p.S101S Chr.4: 68191256-68191256 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764452807 Chr.4: 68191315-68191315 on Build GRCh38
rs1020994092 Chr.4: 68191401-68191401 on Build GRCh38
rs567807204 Chr.4: 68191362-68191362 on Build GRCh38
rs753467165 Chr.4: 68191402-68191402 on Build GRCh38
rs906724979 Chr.4: 68191280-68191280 on Build GRCh38
rs144301451 Chr.4: 68191273-68191273 on Build GRCh38
rs1013878093 Chr.4: 68191268-68191268 on Build GRCh38
rs1000507742 Chr.4: 68191308-68191308 on Build GRCh38
rs751965735 Chr.4: 68191325-68191325 on Build GRCh38
rs754587424 Chr.4: 68191407-68191407 on Build GRCh38
rs533027711 Chr.4: 68191283-68191283 on Build GRCh38
rs951164049 Chr.4: 68191225-68191225 on Build GRCh38
rs536695949 Chr.4: 68191386-68191386 on Build GRCh38
rs4860284 Chr.4: 68191355-68191355 on Build GRCh38
rs370555471 Chr.4: 68191267-68191267 on Build GRCh38
rs762466312 Chr.4: 68191346-68191346 on Build GRCh38
rs183591149 Chr.4: 68191253-68191253 on Build GRCh38
rs535191519 Chr.4: 68191318-68191318 on Build GRCh38
rs771084152 Chr.4: 68191355-68191356 on Build GRCh38
rs1046610918 Chr.4: 68191274-68191274 on Build GRCh38
rs1037700296 Chr.4: 68191260-68191260 on Build GRCh38
rs1052173455 Chr.4: 68191337-68191337 on Build GRCh38
rs749602481 Chr.4: 68191388-68191388 on Build GRCh38
rs114552880 Chr.4: 68191256-68191256 on Build GRCh38
rs749990772 Chr.4: 68191358-68191358 on Build GRCh38
rs551554040 Chr.4: 68191230-68191230 on Build GRCh38
rs1044822151 Chr.4: 68191272-68191272 on Build GRCh38

View Full Product Details