Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 112539732-112539971 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ZGRF1
Entrez Gene ID
55345
Gene Name
zinc finger GRF-type containing 1
Gene Aliases
C4orf21
UniGene
Hs.380346
Gene Chromosome Location
Chr.4: 112539333-112636995 on Build GRCh38
Transcripts
NM_018392.4 XM_005263115.3 XM_011532091.2 XM_011532092.2 XM_011532093.2 XM_011532094.2 XM_011532097.2 XM_011532098.2 XM_011532099.2 XM_017008369.1 XM_017008370.1 XM_017008371.1 XM_017008372.1 XM_017008373.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4150417 c.6077C>A p.A2026E Chr.4: 112539958-112539958 on Build GRCh38
5511360 c.6063_6064insA p.R2022fs*8 Chr.4: 112539971-112539972 on Build GRCh38
4667764 c.6094A>G p.R2032G Chr.4: 112539941-112539941 on Build GRCh38
3824973 c.6105G>A p.L2035L Chr.4: 112539930-112539930 on Build GRCh38
1426404 c.6064_6065insA p.R2022fs*8 Chr.4: 112539970-112539971 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs372375109 Chr.4: 112539908-112539908 on Build GRCh38
rs142786578 Chr.4: 112539862-112539862 on Build GRCh38
rs762755855 Chr.4: 112539882-112539882 on Build GRCh38
rs375291009 Chr.4: 112539917-112539917 on Build GRCh38
rs932986797 Chr.4: 112539830-112539830 on Build GRCh38
rs923034957 Chr.4: 112539791-112539791 on Build GRCh38
rs766570015 Chr.4: 112539853-112539853 on Build GRCh38
rs965483930 Chr.4: 112539869-112539869 on Build GRCh38
rs747819863 Chr.4: 112539927-112539927 on Build GRCh38
rs151025443 Chr.4: 112539864-112539864 on Build GRCh38
rs956715806 Chr.4: 112539780-112539780 on Build GRCh38
rs2306775 Chr.4: 112539771-112539771 on Build GRCh38
rs757706637 Chr.4: 112539894-112539894 on Build GRCh38
rs772996794 Chr.4: 112539854-112539855 on Build GRCh38
rs988260683 Chr.4: 112539810-112539810 on Build GRCh38
rs754351597 Chr.4: 112539890-112539890 on Build GRCh38
rs553097343 Chr.4: 112539816-112539816 on Build GRCh38
rs1032182897 Chr.4: 112539777-112539777 on Build GRCh38
rs767674015 Chr.4: 112539872-112539872 on Build GRCh38
rs764441673 Chr.4: 112539889-112539889 on Build GRCh38
rs541973003 Chr.4: 112539873-112539873 on Build GRCh38
rs746460433 Chr.4: 112539901-112539901 on Build GRCh38
rs147737548 Chr.4: 112539878-112539878 on Build GRCh38
rs759834633 Chr.4: 112539867-112539867 on Build GRCh38

View Full Product Details