Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 7879951-7880176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMP6
Entrez Gene ID
654
Gene Name
bone morphogenetic protein 6
Gene Aliases
VGR, VGR1
UniGene
Hs.285671
Gene Chromosome Location
Chr.6: 7726778-7881728 on Build GRCh38
Transcripts
NM_001718.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4980689 c.1297C>T p.P433S Chr.6: 7880006-7880006 on Build GRCh38
3630077 c.1371C>A p.N457K Chr.6: 7880080-7880080 on Build GRCh38
3630078 c.1377G>A p.A459A Chr.6: 7880086-7880086 on Build GRCh38
5651916 c.1305C>A p.G435G Chr.6: 7880014-7880014 on Build GRCh38
1184774 c.1351G>A p.A451T Chr.6: 7880060-7880060 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs768520334 Chr.6: 7880086-7880086 on Build GRCh38
rs757772931 Chr.6: 7880142-7880142 on Build GRCh38
rs377711135 Chr.6: 7880060-7880060 on Build GRCh38
rs374831278 Chr.6: 7880127-7880127 on Build GRCh38
rs142355450 Chr.6: 7880092-7880092 on Build GRCh38
rs779536486 Chr.6: 7880118-7880119 on Build GRCh38
rs761518530 Chr.6: 7879981-7879981 on Build GRCh38
rs571649629 Chr.6: 7879993-7879993 on Build GRCh38
rs566611196 Chr.6: 7880118-7880122 on Build GRCh38
rs772298426 Chr.6: 7880062-7880062 on Build GRCh38
rs779365354 Chr.6: 7880059-7880059 on Build GRCh38
rs764574165 Chr.6: 7880135-7880135 on Build GRCh38
rs755295884 Chr.6: 7880112-7880113 on Build GRCh38
rs765027412 Chr.6: 7880111-7880111 on Build GRCh38
rs764836345 Chr.6: 7880008-7880008 on Build GRCh38
rs779452476 Chr.6: 7880151-7880151 on Build GRCh38
rs539772264 Chr.6: 7880071-7880071 on Build GRCh38
rs895160763 Chr.6: 7879991-7879991 on Build GRCh38
rs749708761 Chr.6: 7880056-7880056 on Build GRCh38
rs370468169 Chr.6: 7880112-7880112 on Build GRCh38
rs756601097 Chr.6: 7880024-7880024 on Build GRCh38
rs138326174 Chr.6: 7880083-7880083 on Build GRCh38
rs1026161189 Chr.6: 7880034-7880034 on Build GRCh38
rs1013584014 Chr.6: 7880036-7880036 on Build GRCh38
rs150397946 Chr.6: 7880058-7880058 on Build GRCh38
rs766004146 Chr.6: 7880126-7880126 on Build GRCh38
rs761657233 Chr.6: 7880098-7880098 on Build GRCh38

View Full Product Details