Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 4574859-4575084 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf96
Entrez Gene ID
342346
Gene Name
chromosome 16 open reading frame 96
Gene Aliases
-
UniGene
Hs.684212
Gene Chromosome Location
Chr.16: 4538709-4600455 on Build GRCh38
Transcripts
NM_001145011.1 XM_005255298.3 XM_005255301.3 XM_011522475.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6574017 c.607-8C>G p.? Chr.16: 4574964-4574964 on Build GRCh38
6522548 c.658C>A p.L220I Chr.16: 4575023-4575023 on Build GRCh38
4846395 c.690C>T p.T230T Chr.16: 4575055-4575055 on Build GRCh38
4553980 c.607G>A p.A203T Chr.16: 4574972-4574972 on Build GRCh38
4555673 c.663G>A p.W221* Chr.16: 4575028-4575028 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1023286746 Chr.16: 4574900-4574900 on Build GRCh38
rs113353601 Chr.16: 4574936-4574936 on Build GRCh38
rs558135189 Chr.16: 4574930-4574930 on Build GRCh38
rs977527216 Chr.16: 4575017-4575017 on Build GRCh38
rs182165251 Chr.16: 4575041-4575041 on Build GRCh38
rs975850437 Chr.16: 4575010-4575010 on Build GRCh38
rs956949197 Chr.16: 4574918-4574918 on Build GRCh38
rs1026265919 Chr.16: 4574954-4574954 on Build GRCh38
rs766128669 Chr.16: 4575019-4575019 on Build GRCh38
rs935168016 Chr.16: 4575046-4575046 on Build GRCh38
rs775746017 Chr.16: 4574945-4574945 on Build GRCh38
rs549479446 Chr.16: 4575011-4575011 on Build GRCh38
rs964235535 Chr.16: 4575003-4575003 on Build GRCh38
rs567810633 Chr.16: 4574955-4574955 on Build GRCh38
rs1019843177 Chr.16: 4575001-4575001 on Build GRCh38
rs62037460 Chr.16: 4574976-4574976 on Build GRCh38
rs138390932 Chr.16: 4574939-4574939 on Build GRCh38
rs951558791 Chr.16: 4574965-4574965 on Build GRCh38
rs376298041 Chr.16: 4574949-4574949 on Build GRCh38
rs921369497 Chr.16: 4574962-4574962 on Build GRCh38
rs9922925 Chr.16: 4574891-4574891 on Build GRCh38
rs896380268 Chr.16: 4574881-4574881 on Build GRCh38
rs975878059 Chr.16: 4574958-4574958 on Build GRCh38
rs988386388 Chr.16: 4574935-4574935 on Build GRCh38
rs771124202 Chr.16: 4574929-4574929 on Build GRCh38
rs552114198 Chr.16: 4574899-4574899 on Build GRCh38
rs751158306 Chr.16: 4575028-4575028 on Build GRCh38
rs112229339 Chr.16: 4575016-4575016 on Build GRCh38
rs531755391 Chr.16: 4574878-4574878 on Build GRCh38

View Full Product Details