Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 70044271-70044534 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AWAT2
Entrez Gene ID
158835
Gene Name
acyl-CoA wax alcohol acyltransferase 2
Gene Aliases
ARAT, DC4, DGAT2L4, MFAT, WS
UniGene
Hs.661507
Gene Chromosome Location
Chr.X: 70040542-70049970 on Build GRCh38
Transcripts
NM_001002254.1 XM_011530876.2

SNP Detail

SNP ID Chromosome Location
rs773048241 Chr.X: 70044414-70044414 on Build GRCh38
rs747003531 Chr.X: 70044400-70044400 on Build GRCh38
rs894486482 Chr.X: 70044303-70044303 on Build GRCh38
rs768594033 Chr.X: 70044425-70044425 on Build GRCh38
rs771576222 Chr.X: 70044410-70044410 on Build GRCh38
rs757667716 Chr.X: 70044355-70044355 on Build GRCh38
rs758480879 Chr.X: 70044331-70044331 on Build GRCh38
rs765295433 Chr.X: 70044313-70044313 on Build GRCh38
rs766105601 Chr.X: 70044465-70044465 on Build GRCh38
rs1006195235 Chr.X: 70044304-70044304 on Build GRCh38
rs942373560 Chr.X: 70044491-70044491 on Build GRCh38
rs775389096 Chr.X: 70044474-70044474 on Build GRCh38
rs752833138 Chr.X: 70044325-70044325 on Build GRCh38
rs768667252 Chr.X: 70044337-70044338 on Build GRCh38
rs750289991 Chr.X: 70044344-70044344 on Build GRCh38
rs746033647 Chr.X: 70044500-70044511 on Build GRCh38
rs746773340 Chr.X: 70044431-70044431 on Build GRCh38
rs1016384049 Chr.X: 70044342-70044342 on Build GRCh38
rs754120703 Chr.X: 70044493-70044493 on Build GRCh38
rs776698654 Chr.X: 70044440-70044440 on Build GRCh38
rs778221719 Chr.X: 70044403-70044403 on Build GRCh38
rs751868800 Chr.X: 70044354-70044354 on Build GRCh38
rs368039192 Chr.X: 70044490-70044490 on Build GRCh38
rs972263197 Chr.X: 70044381-70044381 on Build GRCh38
rs781144037 Chr.X: 70044441-70044442 on Build GRCh38
rs770921052 Chr.X: 70044434-70044434 on Build GRCh38
rs781641890 Chr.X: 70044391-70044391 on Build GRCh38
rs186195985 Chr.X: 70044450-70044450 on Build GRCh38
rs747727739 Chr.X: 70044404-70044404 on Build GRCh38

View Full Product Details