Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 37377782-37378055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ACACA
Entrez Gene ID
31
Gene Name
acetyl-CoA carboxylase alpha
Gene Aliases
ACAC, ACACAD, ACC, ACC1, ACCA
UniGene
Hs.160556
Gene Chromosome Location
Chr.17: 37084992-37406822 on Build GRCh38
Transcripts
NM_198834.2 NM_198839.2 XM_006721853.1 XM_011524701.1 XM_017024553.1 XM_017024554.1 NM_001321399.1 NM_173625.4 XM_011524647.2 XM_011524648.2 XM_011524649.2

Gene Symbol
C17orf78
Entrez Gene ID
284099
Gene Name
chromosome 17 open reading frame 78
Gene Aliases
-
UniGene
Hs.439154
Gene Chromosome Location
Chr.17: 37375710-37392708 on Build GRCh38
Transcripts
NM_198834.2 NM_198839.2 XM_006721853.1 XM_011524701.1 XM_017024553.1 XM_017024554.1 NM_001321399.1 NM_173625.4 XM_011524647.2 XM_011524648.2 XM_011524649.2

SNP Detail

SNP ID Chromosome Location
rs762910144 Chr.17: 37377895-37377895 on Build GRCh38
rs878932002 Chr.17: 37377909-37377909 on Build GRCh38
rs866629156 Chr.17: 37377936-37377936 on Build GRCh38
rs774112283 Chr.17: 37377900-37377900 on Build GRCh38
rs755649634 Chr.17: 37377843-37377843 on Build GRCh38
rs755516172 Chr.17: 37377948-37377948 on Build GRCh38
rs142727216 Chr.17: 37377996-37377996 on Build GRCh38
rs779601442 Chr.17: 37377951-37377951 on Build GRCh38
rs377481552 Chr.17: 37377978-37377978 on Build GRCh38
rs563867244 Chr.17: 37377925-37377925 on Build GRCh38
rs201523734 Chr.17: 37377846-37377846 on Build GRCh38
rs761638796 Chr.17: 37377911-37377911 on Build GRCh38
rs200503764 Chr.17: 37377876-37377876 on Build GRCh38
rs369234656 Chr.17: 37377899-37377899 on Build GRCh38
rs534868477 Chr.17: 37378005-37378005 on Build GRCh38
rs550347470 Chr.17: 37377888-37377888 on Build GRCh38
rs1050872322 Chr.17: 37377816-37377816 on Build GRCh38
rs771002616 Chr.17: 37377984-37377984 on Build GRCh38
rs373844455 Chr.17: 37377973-37377973 on Build GRCh38
rs35614531 Chr.17: 37377974-37377975 on Build GRCh38
rs377038007 Chr.17: 37377976-37377976 on Build GRCh38
rs371795270 Chr.17: 37377934-37377934 on Build GRCh38
rs112181268 Chr.17: 37378002-37378002 on Build GRCh38
rs990375543 Chr.17: 37377860-37377860 on Build GRCh38
rs367661724 Chr.17: 37377818-37377818 on Build GRCh38
rs551055529 Chr.17: 37377847-37377847 on Build GRCh38
rs76536479 Chr.17: 37377871-37377871 on Build GRCh38
rs748569828 Chr.17: 37377870-37377870 on Build GRCh38
rs549420465 Chr.17: 37377994-37377994 on Build GRCh38
rs374933852 Chr.17: 37377892-37377892 on Build GRCh38
rs781272610 Chr.17: 37377991-37377991 on Build GRCh38
rs977670801 Chr.17: 37377844-37377844 on Build GRCh38
rs766101026 Chr.17: 37378014-37378014 on Build GRCh38
rs747097384 Chr.17: 37377872-37377872 on Build GRCh38
rs779438195 Chr.17: 37377862-37377862 on Build GRCh38
rs570759104 Chr.17: 37378004-37378004 on Build GRCh38
rs754344743 Chr.17: 37377944-37377944 on Build GRCh38
rs115769878 Chr.17: 37377943-37377943 on Build GRCh38
rs552535331 Chr.17: 37377904-37377904 on Build GRCh38
rs745997340 Chr.17: 37377992-37377992 on Build GRCh38
rs184660990 Chr.17: 37377935-37377935 on Build GRCh38
rs900218919 Chr.17: 37377958-37377958 on Build GRCh38
rs372714109 Chr.17: 37377977-37377977 on Build GRCh38

View Full Product Details