Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 103064719-103064952 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V1C1
Entrez Gene ID
528
Gene Name
ATPase H+ transporting V1 subunit C1
Gene Aliases
ATP6C, ATP6D, VATC, Vma5
UniGene
Hs.86905
Gene Chromosome Location
Chr.8: 103021020-103073057 on Build GRCh38
Transcripts
NM_001695.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1094454 c.872T>G p.F291C Chr.8: 103064757-103064757 on Build GRCh38
6608126 c.889G>A p.V297M Chr.8: 103064774-103064774 on Build GRCh38
6444204 c.901C>T p.R301W Chr.8: 103064786-103064786 on Build GRCh38
3395017 c.902G>A p.R301Q Chr.8: 103064787-103064787 on Build GRCh38
7127002 c.909C>A p.F303L Chr.8: 103064794-103064794 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs984057456 Chr.8: 103064808-103064808 on Build GRCh38
rs764064640 Chr.8: 103064748-103064748 on Build GRCh38
rs771591845 Chr.8: 103064859-103064859 on Build GRCh38
rs758123747 Chr.8: 103064794-103064794 on Build GRCh38
rs150964904 Chr.8: 103064795-103064795 on Build GRCh38
rs750239629 Chr.8: 103064774-103064774 on Build GRCh38
rs200782557 Chr.8: 103064823-103064823 on Build GRCh38
rs539308821 Chr.8: 103064838-103064838 on Build GRCh38
rs774963537 Chr.8: 103064860-103064860 on Build GRCh38
rs755973583 Chr.8: 103064862-103064863 on Build GRCh38
rs185652171 Chr.8: 103064913-103064913 on Build GRCh38
rs780539439 Chr.8: 103064848-103064848 on Build GRCh38
rs75246352 Chr.8: 103064919-103064919 on Build GRCh38
rs374255847 Chr.8: 103064787-103064787 on Build GRCh38
rs746235862 Chr.8: 103064863-103064863 on Build GRCh38
rs923247251 Chr.8: 103064891-103064891 on Build GRCh38
rs200373012 Chr.8: 103064786-103064786 on Build GRCh38
rs933408025 Chr.8: 103064898-103064898 on Build GRCh38
rs757109453 Chr.8: 103064764-103064764 on Build GRCh38
rs753878960 Chr.8: 103064755-103064755 on Build GRCh38
rs879209832 Chr.8: 103064926-103064926 on Build GRCh38
rs765066050 Chr.8: 103064773-103064773 on Build GRCh38
rs754584617 Chr.8: 103064839-103064839 on Build GRCh38
rs972086756 Chr.8: 103064881-103064881 on Build GRCh38
rs749858293 Chr.8: 103064856-103064856 on Build GRCh38
rs760820656 Chr.8: 103064740-103064740 on Build GRCh38

View Full Product Details