Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 98822305-98822554 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIAA1211L
Entrez Gene ID
343990
Gene Name
KIAA1211 like
Gene Aliases
C2orf55
UniGene
Hs.469398
Gene Chromosome Location
Chr.2: 98793846-98936221 on Build GRCh38
Transcripts
NM_207362.2 XM_005263923.3 XM_005263924.3 XM_005263926.3 XM_011511093.1 XM_011511094.1 XM_011511095.2 XM_011511096.1 XM_017003971.1 XM_017003972.1 XM_017003973.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5708375 c.1931C>T p.A644V Chr.2: 98822342-98822342 on Build GRCh38
1718243 c.1815_1816GG>AA p.V606I Chr.2: 98822457-98822458 on Build GRCh38
6542873 c.1742C>A p.A581E Chr.2: 98822531-98822531 on Build GRCh38
5952310 c.1800C>T p.L600L Chr.2: 98822473-98822473 on Build GRCh38
6993565 c.1777C>T p.R593W Chr.2: 98822496-98822496 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762228614 Chr.2: 98822525-98822525 on Build GRCh38
rs998540620 Chr.2: 98822335-98822335 on Build GRCh38
rs748050292 Chr.2: 98822324-98822324 on Build GRCh38
rs962470437 Chr.2: 98822385-98822385 on Build GRCh38
rs1029563600 Chr.2: 98822350-98822350 on Build GRCh38
rs928228749 Chr.2: 98822491-98822491 on Build GRCh38
rs772666598 Chr.2: 98822342-98822342 on Build GRCh38
rs751889476 Chr.2: 98822523-98822523 on Build GRCh38
rs770753823 Chr.2: 98822401-98822401 on Build GRCh38
rs866880703 Chr.2: 98822439-98822439 on Build GRCh38
rs935545816 Chr.2: 98822510-98822510 on Build GRCh38
rs759223431 Chr.2: 98822461-98822461 on Build GRCh38
rs916778359 Chr.2: 98822458-98822458 on Build GRCh38
rs771981321 Chr.2: 98822341-98822341 on Build GRCh38
rs941145501 Chr.2: 98822327-98822327 on Build GRCh38
rs982461091 Chr.2: 98822376-98822376 on Build GRCh38
rs867742750 Chr.2: 98822436-98822436 on Build GRCh38
rs991208640 Chr.2: 98822397-98822397 on Build GRCh38
rs1016576869 Chr.2: 98822378-98822378 on Build GRCh38
rs764384247 Chr.2: 98822489-98822489 on Build GRCh38
rs760247806 Chr.2: 98822387-98822387 on Build GRCh38
rs537062302 Chr.2: 98822333-98822333 on Build GRCh38
rs951161079 Chr.2: 98822375-98822375 on Build GRCh38
rs982809685 Chr.2: 98822476-98822476 on Build GRCh38
rs768061978 Chr.2: 98822534-98822534 on Build GRCh38
rs776392242 Chr.2: 98822421-98822421 on Build GRCh38
rs1038590192 Chr.2: 98822339-98822339 on Build GRCh38
rs1010904754 Chr.2: 98822323-98822323 on Build GRCh38
rs915221644 Chr.2: 98822533-98822533 on Build GRCh38

View Full Product Details