Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 144527637-144527864 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C8orf82
Entrez Gene ID
414919
Gene Name
chromosome 8 open reading frame 82
Gene Aliases
-
UniGene
Hs.105685
Gene Chromosome Location
Chr.8: 144526219-144529074 on Build GRCh38
Transcripts
NM_001001795.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5795578 c.206-7C>T p.? Chr.8: 144527794-144527794 on Build GRCh38
6616754 c.349C>A p.L117M Chr.8: 144527644-144527644 on Build GRCh38
4646819 c.329G>T p.R110L Chr.8: 144527664-144527664 on Build GRCh38
7298354 c.209C>T p.P70L Chr.8: 144527784-144527784 on Build GRCh38
5998215 c.336G>A p.V112V Chr.8: 144527657-144527657 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772587199 Chr.8: 144527761-144527761 on Build GRCh38
rs886187981 Chr.8: 144527725-144527725 on Build GRCh38
rs775142920 Chr.8: 144527821-144527821 on Build GRCh38
rs762407196 Chr.8: 144527688-144527688 on Build GRCh38
rs200173614 Chr.8: 144527729-144527729 on Build GRCh38
rs764721650 Chr.8: 144527761-144527763 on Build GRCh38
rs758397404 Chr.8: 144527723-144527723 on Build GRCh38
rs747488205 Chr.8: 144527811-144527811 on Build GRCh38
rs761438375 Chr.8: 144527834-144527834 on Build GRCh38
rs900142713 Chr.8: 144527691-144527691 on Build GRCh38
rs762783867 Chr.8: 144527784-144527784 on Build GRCh38
rs151201677 Chr.8: 144527740-144527740 on Build GRCh38
rs774939404 Chr.8: 144527675-144527675 on Build GRCh38
rs141953984 Chr.8: 144527776-144527776 on Build GRCh38
rs760225997 Chr.8: 144527706-144527706 on Build GRCh38
rs775983976 Chr.8: 144527657-144527657 on Build GRCh38
rs921274208 Chr.8: 144527681-144527681 on Build GRCh38
rs753628038 Chr.8: 144527710-144527710 on Build GRCh38
rs748605285 Chr.8: 144527805-144527805 on Build GRCh38
rs140367665 Chr.8: 144527748-144527748 on Build GRCh38
rs769490617 Chr.8: 144527820-144527820 on Build GRCh38
rs112570002 Chr.8: 144527739-144527739 on Build GRCh38
rs1014434738 Chr.8: 144527768-144527768 on Build GRCh38
rs550232031 Chr.8: 144527664-144527664 on Build GRCh38
rs1036858239 Chr.8: 144527837-144527837 on Build GRCh38
rs578236909 Chr.8: 144527800-144527800 on Build GRCh38
rs780301857 Chr.8: 144527794-144527794 on Build GRCh38
rs940105256 Chr.8: 144527828-144527828 on Build GRCh38
rs375017707 Chr.8: 144527721-144527721 on Build GRCh38
rs763809471 Chr.8: 144527838-144527838 on Build GRCh38
rs762740647 Chr.8: 144527826-144527826 on Build GRCh38
rs748571849 Chr.8: 144527753-144527753 on Build GRCh38
rs772684382 Chr.8: 144527756-144527756 on Build GRCh38
rs201368360 Chr.8: 144527692-144527692 on Build GRCh38
rs766117384 Chr.8: 144527708-144527708 on Build GRCh38
rs781105650 Chr.8: 144527724-144527724 on Build GRCh38
rs202090473 Chr.8: 144527796-144527796 on Build GRCh38
rs764156854 Chr.8: 144527785-144527785 on Build GRCh38
rs145659815 Chr.8: 144527777-144527777 on Build GRCh38
rs756160885 Chr.8: 144527790-144527790 on Build GRCh38
rs749533659 Chr.8: 144527743-144527743 on Build GRCh38
rs756286029 Chr.8: 144527737-144527737 on Build GRCh38
rs960302203 Chr.8: 144527765-144527765 on Build GRCh38
rs990202314 Chr.8: 144527676-144527676 on Build GRCh38
rs751454193 Chr.8: 144527788-144527788 on Build GRCh38
rs538441298 Chr.8: 144527716-144527716 on Build GRCh38
rs894804072 Chr.8: 144527728-144527728 on Build GRCh38
rs912670986 Chr.8: 144527813-144527813 on Build GRCh38
rs769144680 Chr.8: 144527665-144527665 on Build GRCh38
rs138152083 Chr.8: 144527771-144527771 on Build GRCh38
rs751464036 Chr.8: 144527841-144527841 on Build GRCh38
rs939887612 Chr.8: 144527703-144527703 on Build GRCh38
rs954198727 Chr.8: 144527786-144527786 on Build GRCh38
rs778279873 Chr.8: 144527808-144527808 on Build GRCh38
rs772819100 Chr.8: 144527701-144527701 on Build GRCh38
rs371986844 Chr.8: 144527783-144527783 on Build GRCh38
rs371285020 Chr.8: 144527714-144527714 on Build GRCh38
rs371077451 Chr.8: 144527694-144527694 on Build GRCh38
rs370115795 Chr.8: 144527806-144527806 on Build GRCh38
rs367880932 Chr.8: 144527684-144527684 on Build GRCh38
rs776154518 Chr.8: 144527819-144527819 on Build GRCh38
rs754703761 Chr.8: 144527712-144527712 on Build GRCh38
rs554366975 Chr.8: 144527816-144527816 on Build GRCh38
rs969996950 Chr.8: 144527774-144527774 on Build GRCh38
rs912618551 Chr.8: 144527702-144527702 on Build GRCh38
rs376044019 Chr.8: 144527817-144527817 on Build GRCh38
rs558600598 Chr.8: 144527797-144527797 on Build GRCh38
rs369151282 Chr.8: 144527766-144527766 on Build GRCh38
rs745792384 Chr.8: 144527846-144527846 on Build GRCh38
rs752045181 Chr.8: 144527776-144527776 on Build GRCh38
rs554293715 Chr.8: 144527751-144527751 on Build GRCh38
rs919760053 Chr.8: 144527840-144527840 on Build GRCh38

View Full Product Details