Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 15586658-15586925 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C16orf45
Entrez Gene ID
89927
Gene Name
chromosome 16 open reading frame 45
Gene Aliases
MINP
UniGene
Hs.401798
Gene Chromosome Location
Chr.16: 15434468-15594521 on Build GRCh38
Transcripts
NM_001142469.1 NM_033201.2 XM_011522731.2 XM_017023824.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4551114 c.468G>A p.K156K Chr.16: 15586733-15586733 on Build GRCh38
6614483 c.549G>A p.T183T Chr.16: 15586763-15586763 on Build GRCh38
5706015 c.518C>T p.A173V Chr.16: 15586783-15586783 on Build GRCh38
7234198 c.512G>T p.G171V Chr.16: 15586777-15586777 on Build GRCh38
4256315 c.510G>A p.T170T Chr.16: 15586775-15586775 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2071332 Chr.16: 15586785-15586785 on Build GRCh38
rs376264564 Chr.16: 15586790-15586790 on Build GRCh38
rs771341495 Chr.16: 15586860-15586861 on Build GRCh38
rs772197616 Chr.16: 15586703-15586703 on Build GRCh38
rs771465072 Chr.16: 15586689-15586689 on Build GRCh38
rs752334198 Chr.16: 15586791-15586791 on Build GRCh38
rs749430245 Chr.16: 15586829-15586830 on Build GRCh38
rs775574264 Chr.16: 15586706-15586706 on Build GRCh38
rs751373567 Chr.16: 15586723-15586723 on Build GRCh38
rs940010003 Chr.16: 15586861-15586861 on Build GRCh38
rs373862409 Chr.16: 15586762-15586762 on Build GRCh38
rs772506241 Chr.16: 15586869-15586870 on Build GRCh38
rs181574183 Chr.16: 15586707-15586707 on Build GRCh38
rs752246548 Chr.16: 15586728-15586728 on Build GRCh38
rs147145081 Chr.16: 15586837-15586837 on Build GRCh38
rs143456558 Chr.16: 15586802-15586802 on Build GRCh38
rs908551625 Chr.16: 15586847-15586847 on Build GRCh38
rs766316771 Chr.16: 15586787-15586787 on Build GRCh38
rs757776734 Chr.16: 15586813-15586813 on Build GRCh38
rs771121939 Chr.16: 15586901-15586901 on Build GRCh38
rs748403294 Chr.16: 15586725-15586736 on Build GRCh38
rs200144830 Chr.16: 15586870-15586870 on Build GRCh38
rs760370740 Chr.16: 15586874-15586878 on Build GRCh38
rs775378313 Chr.16: 15586869-15586869 on Build GRCh38
rs775239843 Chr.16: 15586874-15586876 on Build GRCh38
rs142995376 Chr.16: 15586726-15586726 on Build GRCh38
rs746234191 Chr.16: 15586822-15586822 on Build GRCh38
rs753412404 Chr.16: 15586810-15586810 on Build GRCh38
rs370378624 Chr.16: 15586823-15586823 on Build GRCh38
rs368962560 Chr.16: 15586721-15586721 on Build GRCh38
rs113251387 Chr.16: 15586716-15586716 on Build GRCh38
rs938191154 Chr.16: 15586896-15586896 on Build GRCh38
rs35954715 Chr.16: 15586861-15586861 on Build GRCh38
rs776720007 Chr.16: 15586775-15586775 on Build GRCh38
rs756968047 Chr.16: 15586693-15586693 on Build GRCh38
rs578226555 Chr.16: 15586752-15586752 on Build GRCh38
rs748229986 Chr.16: 15586863-15586863 on Build GRCh38
rs777811579 Chr.16: 15586790-15586790 on Build GRCh38
rs137927249 Chr.16: 15586722-15586722 on Build GRCh38
rs750407106 Chr.16: 15586718-15586718 on Build GRCh38
rs778394779 Chr.16: 15586742-15586742 on Build GRCh38
rs770044485 Chr.16: 15586742-15586742 on Build GRCh38
rs753398131 Chr.16: 15586881-15586881 on Build GRCh38
rs763650544 Chr.16: 15586803-15586803 on Build GRCh38
rs1127294 Chr.16: 15586900-15586900 on Build GRCh38
rs371793732 Chr.16: 15586697-15586697 on Build GRCh38
rs759350784 Chr.16: 15586868-15586868 on Build GRCh38
rs759757395 Chr.16: 15586861-15586862 on Build GRCh38
rs763843351 Chr.16: 15586870-15586870 on Build GRCh38
rs149422405 Chr.16: 15586774-15586774 on Build GRCh38
rs138950756 Chr.16: 15586766-15586766 on Build GRCh38
rs765210216 Chr.16: 15586709-15586709 on Build GRCh38
rs749718364 Chr.16: 15586875-15586875 on Build GRCh38
rs919987473 Chr.16: 15586890-15586890 on Build GRCh38
rs751287668 Chr.16: 15586788-15586788 on Build GRCh38
rs561734612 Chr.16: 15586821-15586821 on Build GRCh38
rs368775133 Chr.16: 15586694-15586694 on Build GRCh38
rs150243763 Chr.16: 15586763-15586763 on Build GRCh38
rs747156942 Chr.16: 15586830-15586830 on Build GRCh38
rs755635266 Chr.16: 15586733-15586733 on Build GRCh38
rs768885184 Chr.16: 15586836-15586836 on Build GRCh38
rs763886253 Chr.16: 15586876-15586877 on Build GRCh38
rs780290225 Chr.16: 15586826-15586826 on Build GRCh38
rs749773994 Chr.16: 15586750-15586750 on Build GRCh38
rs748553314 Chr.16: 15586779-15586779 on Build GRCh38
rs779363288 Chr.16: 15586757-15586757 on Build GRCh38
rs902512787 Chr.16: 15586687-15586687 on Build GRCh38
rs774212028 Chr.16: 15586867-15586867 on Build GRCh38

View Full Product Details