Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 69449415-69449680 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
APBA1
Entrez Gene ID
320
Gene Name
amyloid beta precursor protein binding family A member 1
Gene Aliases
D9S411E, LIN10, MINT1, X11, X11A, X11ALPHA
UniGene
Hs.171939
Gene Chromosome Location
Chr.9: 69427532-69673012 on Build GRCh38
Transcripts
NM_001163.3 XM_005251968.3 XM_011518617.2 XM_017014670.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1643859 c.2166C>T p.C722C Chr.9: 69449599-69449599 on Build GRCh38
7238511 c.2130T>A p.N710K Chr.9: 69449635-69449635 on Build GRCh38
4775980 c.2132G>C p.G711A Chr.9: 69449633-69449633 on Build GRCh38
3657834 c.2118C>T p.I706I Chr.9: 69449647-69449647 on Build GRCh38
5876332 c.2093G>A p.G698E Chr.9: 69449672-69449672 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764083422 Chr.9: 69449640-69449640 on Build GRCh38
rs374466871 Chr.9: 69449628-69449628 on Build GRCh38
rs1042414801 Chr.9: 69449463-69449463 on Build GRCh38
rs889857005 Chr.9: 69449562-69449562 on Build GRCh38
rs1055286224 Chr.9: 69449450-69449450 on Build GRCh38
rs879162392 Chr.9: 69449534-69449534 on Build GRCh38
rs747451624 Chr.9: 69449564-69449564 on Build GRCh38
rs775678882 Chr.9: 69449558-69449558 on Build GRCh38
rs759268751 Chr.9: 69449614-69449614 on Build GRCh38
rs746113941 Chr.9: 69449538-69449538 on Build GRCh38
rs779023132 Chr.9: 69449519-69449519 on Build GRCh38
rs773829845 Chr.9: 69449603-69449603 on Build GRCh38
rs1006709394 Chr.9: 69449571-69449571 on Build GRCh38
rs946949462 Chr.9: 69449456-69449456 on Build GRCh38
rs915529374 Chr.9: 69449451-69449451 on Build GRCh38
rs572989691 Chr.9: 69449578-69449578 on Build GRCh38
rs762204468 Chr.9: 69449574-69449574 on Build GRCh38
rs985726747 Chr.9: 69449476-69449476 on Build GRCh38
rs140376953 Chr.9: 69449516-69449516 on Build GRCh38
rs866287545 Chr.9: 69449542-69449542 on Build GRCh38
rs377604682 Chr.9: 69449543-69449543 on Build GRCh38
rs949370569 Chr.9: 69449631-69449631 on Build GRCh38
rs556548354 Chr.9: 69449503-69449503 on Build GRCh38
rs962644565 Chr.9: 69449630-69449630 on Build GRCh38
rs765713119 Chr.9: 69449589-69449589 on Build GRCh38
rs753778142 Chr.9: 69449645-69449645 on Build GRCh38
rs199985605 Chr.9: 69449618-69449618 on Build GRCh38
rs766901377 Chr.9: 69449617-69449617 on Build GRCh38
rs998291047 Chr.9: 69449506-69449506 on Build GRCh38
rs144248964 Chr.9: 69449572-69449572 on Build GRCh38
rs942953916 Chr.9: 69449505-69449506 on Build GRCh38

View Full Product Details