Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 114400166-114400415 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C11orf71
Entrez Gene ID
54494
Gene Name
chromosome 11 open reading frame 71
Gene Aliases
URLC7
UniGene
Hs.715083
Gene Chromosome Location
Chr.11: 114391448-114400550 on Build GRCh38
Transcripts
NM_001271562.1 NM_019021.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3868471 c.137C>T p.P46L Chr.11: 114400195-114400195 on Build GRCh38
6444243 c.75C>A p.D25E Chr.11: 114400257-114400257 on Build GRCh38
6552687 c.71G>A p.G24D Chr.11: 114400261-114400261 on Build GRCh38
5635050 c.34G>A p.D12N Chr.11: 114400298-114400298 on Build GRCh38
297932 c.96G>A p.A32A Chr.11: 114400236-114400236 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs371796136 Chr.11: 114400366-114400366 on Build GRCh38
rs566705091 Chr.11: 114400394-114400394 on Build GRCh38
rs898242148 Chr.11: 114400302-114400302 on Build GRCh38
rs369588538 Chr.11: 114400288-114400288 on Build GRCh38
rs9882 Chr.11: 114400243-114400243 on Build GRCh38
rs781557921 Chr.11: 114400256-114400256 on Build GRCh38
rs749900235 Chr.11: 114400234-114400234 on Build GRCh38
rs199945919 Chr.11: 114400307-114400307 on Build GRCh38
rs764740213 Chr.11: 114400231-114400231 on Build GRCh38
rs375198468 Chr.11: 114400217-114400217 on Build GRCh38
rs115808383 Chr.11: 114400275-114400275 on Build GRCh38
rs749838579 Chr.11: 114400354-114400354 on Build GRCh38
rs955945622 Chr.11: 114400316-114400316 on Build GRCh38
rs1032779003 Chr.11: 114400358-114400358 on Build GRCh38
rs377553860 Chr.11: 114400303-114400303 on Build GRCh38
rs570461368 Chr.11: 114400355-114400355 on Build GRCh38
rs779150791 Chr.11: 114400364-114400364 on Build GRCh38
rs980290847 Chr.11: 114400345-114400345 on Build GRCh38
rs773177096 Chr.11: 114400205-114400205 on Build GRCh38
rs768517083 Chr.11: 114400290-114400290 on Build GRCh38
rs188164114 Chr.11: 114400321-114400321 on Build GRCh38
rs748595867 Chr.11: 114400261-114400261 on Build GRCh38
rs770288194 Chr.11: 114400268-114400268 on Build GRCh38
rs920824298 Chr.11: 114400362-114400362 on Build GRCh38
rs772422711 Chr.11: 114400367-114400367 on Build GRCh38
rs944742083 Chr.11: 114400392-114400392 on Build GRCh38
rs886493884 Chr.11: 114400197-114400197 on Build GRCh38
rs761769384 Chr.11: 114400306-114400306 on Build GRCh38
rs192687124 Chr.11: 114400215-114400215 on Build GRCh38
rs550501384 Chr.11: 114400335-114400335 on Build GRCh38
rs957044303 Chr.11: 114400213-114400213 on Build GRCh38
rs181995153 Chr.11: 114400325-114400325 on Build GRCh38
rs200550352 Chr.11: 114400331-114400331 on Build GRCh38
rs754636336 Chr.11: 114400327-114400327 on Build GRCh38
rs369811614 Chr.11: 114400389-114400389 on Build GRCh38
rs781642078 Chr.11: 114400189-114400189 on Build GRCh38
rs373835784 Chr.11: 114400244-114400244 on Build GRCh38
rs553174314 Chr.11: 114400390-114400390 on Build GRCh38
rs779553438 Chr.11: 114400242-114400242 on Build GRCh38
rs762340956 Chr.11: 114400310-114400310 on Build GRCh38
rs1038036891 Chr.11: 114400298-114400298 on Build GRCh38
rs766998280 Chr.11: 114400317-114400317 on Build GRCh38
rs900119686 Chr.11: 114400347-114400347 on Build GRCh38
rs769815897 Chr.11: 114400201-114400201 on Build GRCh38
rs374455783 Chr.11: 114400294-114400294 on Build GRCh38
rs760519922 Chr.11: 114400219-114400219 on Build GRCh38
rs773253208 Chr.11: 114400309-114400309 on Build GRCh38
rs775916604 Chr.11: 114400381-114400381 on Build GRCh38
rs370321807 Chr.11: 114400382-114400382 on Build GRCh38
rs774402019 Chr.11: 114400287-114400287 on Build GRCh38
rs769057261 Chr.11: 114400188-114400188 on Build GRCh38
rs778408255 Chr.11: 114400272-114400272 on Build GRCh38
rs561777897 Chr.11: 114400195-114400195 on Build GRCh38
rs764017168 Chr.11: 114400220-114400220 on Build GRCh38
rs564422718 Chr.11: 114400223-114400223 on Build GRCh38
rs368055793 Chr.11: 114400334-114400334 on Build GRCh38
rs544732282 Chr.11: 114400221-114400221 on Build GRCh38
rs112068263 Chr.11: 114400239-114400239 on Build GRCh38
rs754055816 Chr.11: 114400346-114400346 on Build GRCh38

View Full Product Details