Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 141907070-141907343 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP1B3
Entrez Gene ID
483
Gene Name
ATPase Na+/K+ transporting subunit beta 3
Gene Aliases
ATPB-3, CD298
UniGene
Hs.477789
Gene Chromosome Location
Chr.3: 141876628-141926540 on Build GRCh38
Transcripts

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6226085 c.268G>A p.A90T Chr.3: 141907196-141907196 on Build GRCh38
460898 c.305C>T p.S102L Chr.3: 141907233-141907233 on Build GRCh38
3427160 c.325G>A p.D109N Chr.3: 141907253-141907253 on Build GRCh38
729001 c.328C>A p.L110I Chr.3: 141907256-141907256 on Build GRCh38
5006682 c.260C>T p.P87L Chr.3: 141907188-141907188 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs182080039 Chr.3: 141907144-141907144 on Build GRCh38
rs3792411 Chr.3: 141907281-141907281 on Build GRCh38
rs763767863 Chr.3: 141907318-141907318 on Build GRCh38
rs778948900 Chr.3: 141907199-141907199 on Build GRCh38
rs142238487 Chr.3: 141907131-141907131 on Build GRCh38
rs927531200 Chr.3: 141907106-141907106 on Build GRCh38
rs545520064 Chr.3: 141907149-141907149 on Build GRCh38
rs777316470 Chr.3: 141907234-141907234 on Build GRCh38
rs371578919 Chr.3: 141907316-141907316 on Build GRCh38
rs775513549 Chr.3: 141907218-141907218 on Build GRCh38
rs754665671 Chr.3: 141907152-141907152 on Build GRCh38
rs757370922 Chr.3: 141907198-141907198 on Build GRCh38
rs72988221 Chr.3: 141907195-141907195 on Build GRCh38
rs1047113913 Chr.3: 141907164-141907164 on Build GRCh38
rs772339404 Chr.3: 141907214-141907214 on Build GRCh38
rs746153050 Chr.3: 141907210-141907210 on Build GRCh38
rs777792091 Chr.3: 141907196-141907196 on Build GRCh38
rs770624951 Chr.3: 141907243-141907243 on Build GRCh38
rs7622417 Chr.3: 141907157-141907157 on Build GRCh38
rs368376486 Chr.3: 141907315-141907315 on Build GRCh38
rs747422486 Chr.3: 141907219-141907219 on Build GRCh38
rs751147355 Chr.3: 141907293-141907293 on Build GRCh38
rs886274870 Chr.3: 141907173-141907173 on Build GRCh38
rs370569371 Chr.3: 141907275-141907275 on Build GRCh38
rs377366509 Chr.3: 141907239-141907239 on Build GRCh38
rs995865803 Chr.3: 141907274-141907274 on Build GRCh38
rs74650318 Chr.3: 141907314-141907314 on Build GRCh38
rs752541136 Chr.3: 141907160-141907160 on Build GRCh38
rs776954006 Chr.3: 141907139-141907139 on Build GRCh38
rs559746096 Chr.3: 141907135-141907135 on Build GRCh38
rs751307573 Chr.3: 141907150-141907150 on Build GRCh38
rs768861940 Chr.3: 141907233-141907233 on Build GRCh38

View Full Product Details