Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 114638357-114638584 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM268
Entrez Gene ID
203197
Gene Name
transmembrane protein 268
Gene Aliases
C9orf91
UniGene
Hs.522357
Gene Chromosome Location
Chr.9: 114603897-114646423 on Build GRCh38
Transcripts
NM_153045.3 XM_006716997.1 XM_011518350.1 XM_011518351.2 XM_011518352.1 XM_011518353.1 XM_011518354.1 XM_011518355.1 XM_017014429.1 XM_017014430.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3996260 c.694T>C p.L232L Chr.9: 114638571-114638571 on Build GRCh38
1659698 c.667-3C>A p.? Chr.9: 114638541-114638541 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs767565790 Chr.9: 114638496-114638496 on Build GRCh38
rs749057268 Chr.9: 114638515-114638515 on Build GRCh38
rs778872495 Chr.9: 114638522-114638522 on Build GRCh38
rs752970994 Chr.9: 114638497-114638497 on Build GRCh38
rs891136070 Chr.9: 114638541-114638541 on Build GRCh38
rs1038424812 Chr.9: 114638489-114638489 on Build GRCh38
rs941874339 Chr.9: 114638474-114638474 on Build GRCh38
rs189893326 Chr.9: 114638523-114638523 on Build GRCh38
rs777544003 Chr.9: 114638513-114638513 on Build GRCh38
rs570791505 Chr.9: 114638407-114638407 on Build GRCh38
rs542553995 Chr.9: 114638456-114638456 on Build GRCh38
rs778031912 Chr.9: 114638535-114638535 on Build GRCh38
rs542041191 Chr.9: 114638549-114638549 on Build GRCh38
rs754408152 Chr.9: 114638390-114638390 on Build GRCh38
rs757168475 Chr.9: 114638516-114638516 on Build GRCh38
rs949313785 Chr.9: 114638425-114638425 on Build GRCh38
rs773470330 Chr.9: 114638562-114638562 on Build GRCh38
rs1036528198 Chr.9: 114638524-114638524 on Build GRCh38
rs886231673 Chr.9: 114638436-114638436 on Build GRCh38
rs756321292 Chr.9: 114638510-114638510 on Build GRCh38
rs775127409 Chr.9: 114638539-114638539 on Build GRCh38
rs34263982 Chr.9: 114638398-114638399 on Build GRCh38
rs755371404 Chr.9: 114638490-114638490 on Build GRCh38
rs147417909 Chr.9: 114638431-114638431 on Build GRCh38
rs145315370 Chr.9: 114638546-114638546 on Build GRCh38

View Full Product Details