Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 70125996-70126269 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANO1
Entrez Gene ID
55107
Gene Name
anoctamin 1
Gene Aliases
DOG1, ORAOV2, TAOS2, TMEM16A
UniGene
Hs.503074
Gene Chromosome Location
Chr.11: 70078302-70189545 on Build GRCh38
Transcripts
NM_018043.5 NR_030691.1 XM_006718602.2 XM_006718604.2 XM_006718605.2 XM_011545121.2 XM_011545123.2 XM_011545124.2 XM_011545125.2 XM_011545126.2 XM_011545127.2 XM_011545128.2 XM_011545129.2 XM_011545131.2 XM_017017956.1 XM_017017957.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
118542 c.1068A>T p.G356G Chr.11: 70126166-70126166 on Build GRCh38
6134369 c.997G>C p.A333P Chr.11: 70126095-70126095 on Build GRCh38
4775322 c.931A>C p.T311P Chr.11: 70126113-70126113 on Build GRCh38
3452674 c.1092C>T p.I364I Chr.11: 70126190-70126190 on Build GRCh38
1194053 c.1007G>T p.G336V Chr.11: 70126105-70126105 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201448245 Chr.11: 70126106-70126106 on Build GRCh38
rs546073769 Chr.11: 70126224-70126224 on Build GRCh38
rs572735290 Chr.11: 70126211-70126211 on Build GRCh38
rs772929785 Chr.11: 70126083-70126083 on Build GRCh38
rs757887721 Chr.11: 70126152-70126152 on Build GRCh38
rs761985421 Chr.11: 70126137-70126137 on Build GRCh38
rs929747829 Chr.11: 70126122-70126122 on Build GRCh38
rs764240546 Chr.11: 70126067-70126067 on Build GRCh38
rs774347182 Chr.11: 70126232-70126232 on Build GRCh38
rs376965274 Chr.11: 70126139-70126139 on Build GRCh38
rs745875650 Chr.11: 70126218-70126218 on Build GRCh38
rs778326070 Chr.11: 70126190-70126190 on Build GRCh38
rs767798144 Chr.11: 70126138-70126138 on Build GRCh38
rs542012166 Chr.11: 70126170-70126170 on Build GRCh38
rs760485998 Chr.11: 70126088-70126088 on Build GRCh38
rs780505960 Chr.11: 70126157-70126157 on Build GRCh38
rs1030322617 Chr.11: 70126233-70126233 on Build GRCh38
rs1041394046 Chr.11: 70126231-70126231 on Build GRCh38
rs866069288 Chr.11: 70126129-70126129 on Build GRCh38
rs770727713 Chr.11: 70126212-70126212 on Build GRCh38
rs371709085 Chr.11: 70126049-70126049 on Build GRCh38
rs74943286 Chr.11: 70126150-70126150 on Build GRCh38
rs746598808 Chr.11: 70126205-70126205 on Build GRCh38
rs1039514594 Chr.11: 70126174-70126174 on Build GRCh38
rs570844488 Chr.11: 70126169-70126169 on Build GRCh38
rs751658523 Chr.11: 70126128-70126128 on Build GRCh38
rs761096522 Chr.11: 70126217-70126220 on Build GRCh38
rs761980533 Chr.11: 70126235-70126235 on Build GRCh38
rs560760995 Chr.11: 70126201-70126201 on Build GRCh38
rs776556418 Chr.11: 70126095-70126095 on Build GRCh38
rs373398897 Chr.11: 70126241-70126241 on Build GRCh38
rs757797751 Chr.11: 70126202-70126202 on Build GRCh38
rs200652721 Chr.11: 70126217-70126217 on Build GRCh38
rs201648677 Chr.11: 70126082-70126082 on Build GRCh38
rs768292606 Chr.11: 70126029-70126029 on Build GRCh38
rs775914955 Chr.11: 70126049-70126049 on Build GRCh38
rs757769819 Chr.11: 70126163-70126163 on Build GRCh38
rs747820898 Chr.11: 70126069-70126069 on Build GRCh38
rs764219083 Chr.11: 70126118-70126118 on Build GRCh38
rs368980516 Chr.11: 70126094-70126094 on Build GRCh38

View Full Product Details