Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 5739251-5739516 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CATSPERD
Entrez Gene ID
257062
Gene Name
cation channel sperm associated auxiliary subunit delta
Gene Aliases
TMEM146
UniGene
Hs.631842
Gene Chromosome Location
Chr.19: 5720385-5778734 on Build GRCh38
Transcripts
NM_152784.3 XM_011527885.2 XM_011527886.2 XM_011527887.2 XM_011527888.2 XM_011527889.2 XM_011527890.2 XM_017026564.1 XM_017026565.1 XM_017026566.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3404698 c.475G>C p.V159L Chr.19: 5739341-5739341 on Build GRCh38
6389656 c.482C>T p.A161V Chr.19: 5739348-5739348 on Build GRCh38
4081967 c.500A>G p.Q167R Chr.19: 5739366-5739366 on Build GRCh38
3720343 c.544A>C p.S182R Chr.19: 5739410-5739410 on Build GRCh38
4403093 c.477T>G p.V159V Chr.19: 5739343-5739343 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771051358 Chr.19: 5739349-5739349 on Build GRCh38
rs79705431 Chr.19: 5739322-5739322 on Build GRCh38
rs146087186 Chr.19: 5739480-5739480 on Build GRCh38
rs199838260 Chr.19: 5739440-5739440 on Build GRCh38
rs990795177 Chr.19: 5739395-5739395 on Build GRCh38
rs766736636 Chr.19: 5739314-5739314 on Build GRCh38
rs768354338 Chr.19: 5739310-5739311 on Build GRCh38
rs755633754 Chr.19: 5739306-5739310 on Build GRCh38
rs371002531 Chr.19: 5739356-5739356 on Build GRCh38
rs747953267 Chr.19: 5739310-5739313 on Build GRCh38
rs571512657 Chr.19: 5739493-5739493 on Build GRCh38
rs749272464 Chr.19: 5739339-5739339 on Build GRCh38
rs376258601 Chr.19: 5739309-5739310 on Build GRCh38
rs936210002 Chr.19: 5739394-5739394 on Build GRCh38
rs752093724 Chr.19: 5739321-5739321 on Build GRCh38
rs781514684 Chr.19: 5739332-5739332 on Build GRCh38
rs754118836 Chr.19: 5739477-5739477 on Build GRCh38
rs772928665 Chr.19: 5739382-5739382 on Build GRCh38
rs550005895 Chr.19: 5739310-5739310 on Build GRCh38
rs780097143 Chr.19: 5739352-5739352 on Build GRCh38
rs765898881 Chr.19: 5739407-5739407 on Build GRCh38
rs761733551 Chr.19: 5739365-5739365 on Build GRCh38
rs1005463692 Chr.19: 5739292-5739292 on Build GRCh38
rs200285271 Chr.19: 5739326-5739326 on Build GRCh38
rs199601443 Chr.19: 5739357-5739357 on Build GRCh38
rs753101571 Chr.19: 5739335-5739335 on Build GRCh38
rs373873841 Chr.19: 5739302-5739302 on Build GRCh38
rs764979510 Chr.19: 5739298-5739298 on Build GRCh38
rs756290061 Chr.19: 5739336-5739336 on Build GRCh38
rs762302113 Chr.19: 5739399-5739399 on Build GRCh38
rs879231480 Chr.19: 5739301-5739301 on Build GRCh38
rs756518816 Chr.19: 5739468-5739468 on Build GRCh38
rs745981294 Chr.19: 5739484-5739484 on Build GRCh38
rs142868547 Chr.19: 5739275-5739275 on Build GRCh38
rs751121808 Chr.19: 5739421-5739421 on Build GRCh38
rs564702647 Chr.19: 5739274-5739274 on Build GRCh38
rs753050559 Chr.19: 5739464-5739464 on Build GRCh38
rs764158966 Chr.19: 5739476-5739476 on Build GRCh38
rs757232157 Chr.19: 5739479-5739479 on Build GRCh38
rs745399347 Chr.19: 5739299-5739302 on Build GRCh38
rs768079757 Chr.19: 5739450-5739450 on Build GRCh38
rs1017218477 Chr.19: 5739294-5739294 on Build GRCh38
rs769317730 Chr.19: 5739366-5739366 on Build GRCh38
rs201076963 Chr.19: 5739338-5739338 on Build GRCh38
rs761345749 Chr.19: 5739285-5739285 on Build GRCh38
rs762579398 Chr.19: 5739313-5739313 on Build GRCh38

View Full Product Details