Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 90954136-90954409 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCER1
Entrez Gene ID
196477
Gene Name
coiled-coil glutamate rich protein 1
Gene Aliases
C12orf12
UniGene
Hs.128574
Gene Chromosome Location
Chr.12: 90952215-90955176 on Build GRCh38
Transcripts
NM_152638.3 NR_130711.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3688481 c.410C>T p.P137L Chr.12: 90954333-90954333 on Build GRCh38
6238852 c.414C>T p.G138G Chr.12: 90954329-90954329 on Build GRCh38
4152676 c.443G>A p.R148H Chr.12: 90954300-90954300 on Build GRCh38
352474 c.397T>C p.W133R Chr.12: 90954346-90954346 on Build GRCh38
4878377 c.334T>C p.Y112H Chr.12: 90954409-90954409 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs920509298 Chr.12: 90954308-90954308 on Build GRCh38
rs868406759 Chr.12: 90954321-90954321 on Build GRCh38
rs369758757 Chr.12: 90954298-90954298 on Build GRCh38
rs769288159 Chr.12: 90954209-90954209 on Build GRCh38
rs374385699 Chr.12: 90954234-90954234 on Build GRCh38
rs767881614 Chr.12: 90954160-90954160 on Build GRCh38
rs766714351 Chr.12: 90954355-90954385 on Build GRCh38
rs747767336 Chr.12: 90954299-90954299 on Build GRCh38
rs201558156 Chr.12: 90954280-90954280 on Build GRCh38
rs771361475 Chr.12: 90954228-90954228 on Build GRCh38
rs746803573 Chr.12: 90954183-90954183 on Build GRCh38
rs777284354 Chr.12: 90954309-90954309 on Build GRCh38
rs748776182 Chr.12: 90954319-90954319 on Build GRCh38
rs756279053 Chr.12: 90954170-90954170 on Build GRCh38
rs373379300 Chr.12: 90954282-90954282 on Build GRCh38
rs115451948 Chr.12: 90954222-90954222 on Build GRCh38
rs932656123 Chr.12: 90954229-90954229 on Build GRCh38
rs540718130 Chr.12: 90954161-90954161 on Build GRCh38
rs144591989 Chr.12: 90954283-90954283 on Build GRCh38
rs780834488 Chr.12: 90954205-90954205 on Build GRCh38
rs764331734 Chr.12: 90954171-90954171 on Build GRCh38
rs377744456 Chr.12: 90954252-90954252 on Build GRCh38
rs774734041 Chr.12: 90954231-90954231 on Build GRCh38
rs746448692 Chr.12: 90954344-90954344 on Build GRCh38
rs370019436 Chr.12: 90954214-90954214 on Build GRCh38
rs943880423 Chr.12: 90954207-90954207 on Build GRCh38
rs554003370 Chr.12: 90954304-90954304 on Build GRCh38
rs1014407388 Chr.12: 90954315-90954315 on Build GRCh38
rs1053185578 Chr.12: 90954267-90954267 on Build GRCh38
rs200199546 Chr.12: 90954258-90954258 on Build GRCh38
rs1026570491 Chr.12: 90954346-90954346 on Build GRCh38
rs866986813 Chr.12: 90954318-90954318 on Build GRCh38
rs906086902 Chr.12: 90954335-90954335 on Build GRCh38
rs750375255 Chr.12: 90954174-90954174 on Build GRCh38
rs952380701 Chr.12: 90954358-90954358 on Build GRCh38
rs17852341 Chr.12: 90954281-90954281 on Build GRCh38
rs779878350 Chr.12: 90954180-90954180 on Build GRCh38
rs541956220 Chr.12: 90954337-90954337 on Build GRCh38
rs776041441 Chr.12: 90954378-90954378 on Build GRCh38
rs754621257 Chr.12: 90954193-90954193 on Build GRCh38
rs775845183 Chr.12: 90954237-90954237 on Build GRCh38
rs758307347 Chr.12: 90954279-90954279 on Build GRCh38
rs763512900 Chr.12: 90954192-90954193 on Build GRCh38
rs952433664 Chr.12: 90954176-90954176 on Build GRCh38
rs772564857 Chr.12: 90954372-90954372 on Build GRCh38
rs753914225 Chr.12: 90954263-90954263 on Build GRCh38
rs868224869 Chr.12: 90954331-90954331 on Build GRCh38
rs759820995 Chr.12: 90954233-90954233 on Build GRCh38
rs760957326 Chr.12: 90954380-90954380 on Build GRCh38

View Full Product Details