Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 57745332-57745595 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BTBD18
Entrez Gene ID
643376
Gene Name
BTB domain containing 18
Gene Aliases
-
UniGene
Hs.606355
Gene Chromosome Location
Chr.11: 57743514-57753176 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NR_037646.1

Gene Symbol
TMX2-CTNND1
Entrez Gene ID
100528016
Gene Name
TMX2-CTNND1 readthrough (NMD candidate)
Gene Aliases
-
UniGene
Hs.166011
Gene Chromosome Location
Chr.11: 57712523-57819180 on Build GRCh38
Transcripts
NM_001145101.1 XM_011545212.2 XM_017018127.1 XM_017018128.1 NR_037646.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4977847 c.721G>T p.D241Y Chr.11: 57745552-57745552 on Build GRCh38
1354818 c.912T>C p.N304N Chr.11: 57745361-57745361 on Build GRCh38
5035617 c.809C>A p.P270Q Chr.11: 57745464-57745464 on Build GRCh38
7194378 c.825T>C p.C275C Chr.11: 57745448-57745448 on Build GRCh38
4631501 c.800G>A p.R267H Chr.11: 57745473-57745473 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs368415279 Chr.11: 57745389-57745389 on Build GRCh38
rs12787776 Chr.11: 57745515-57745515 on Build GRCh38
rs749346935 Chr.11: 57745386-57745386 on Build GRCh38
rs1010405978 Chr.11: 57745357-57745357 on Build GRCh38
rs569933884 Chr.11: 57745423-57745423 on Build GRCh38
rs755434207 Chr.11: 57745502-57745502 on Build GRCh38
rs774530773 Chr.11: 57745398-57745398 on Build GRCh38
rs745718258 Chr.11: 57745401-57745401 on Build GRCh38
rs549782162 Chr.11: 57745452-57745452 on Build GRCh38
rs12794985 Chr.11: 57745566-57745566 on Build GRCh38
rs764057238 Chr.11: 57745491-57745491 on Build GRCh38
rs536677168 Chr.11: 57745361-57745361 on Build GRCh38
rs776048962 Chr.11: 57745527-57745527 on Build GRCh38
rs945477818 Chr.11: 57745542-57745542 on Build GRCh38
rs992270250 Chr.11: 57745487-57745487 on Build GRCh38
rs140102739 Chr.11: 57745448-57745448 on Build GRCh38
rs529887501 Chr.11: 57745360-57745360 on Build GRCh38
rs979267290 Chr.11: 57745559-57745559 on Build GRCh38
rs1006957962 Chr.11: 57745387-57745387 on Build GRCh38
rs756584253 Chr.11: 57745366-57745366 on Build GRCh38
rs771179268 Chr.11: 57745390-57745390 on Build GRCh38
rs775300523 Chr.11: 57745446-57745446 on Build GRCh38
rs913936202 Chr.11: 57745541-57745541 on Build GRCh38
rs78162678 Chr.11: 57745368-57745368 on Build GRCh38

View Full Product Details