Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 76134149-76134410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TMEM266
Entrez Gene ID
123591
Gene Name
transmembrane protein 266
Gene Aliases
C15orf27, HVRP1
UniGene
Hs.661411
Gene Chromosome Location
Chr.15: 76059929-76204963 on Build GRCh38
Transcripts
NM_152335.2 XM_005254160.2 XM_017021915.1 XM_017021916.1 XM_017021917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3887519 c.13C>T p.P5S Chr.15: 76134276-76134276 on Build GRCh38
6310405 c.25A>G p.M9V Chr.15: 76134288-76134288 on Build GRCh38
5012916 c.27G>T p.M9I Chr.15: 76134290-76134290 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs757523429 Chr.15: 76134324-76134324 on Build GRCh38
rs188062903 Chr.15: 76134202-76134202 on Build GRCh38
rs753130982 Chr.15: 76134217-76134217 on Build GRCh38
rs779834014 Chr.15: 76134262-76134262 on Build GRCh38
rs769403172 Chr.15: 76134250-76134250 on Build GRCh38
rs780994606 Chr.15: 76134220-76134220 on Build GRCh38
rs760964744 Chr.15: 76134201-76134201 on Build GRCh38
rs958342165 Chr.15: 76134221-76134221 on Build GRCh38
rs889320719 Chr.15: 76134200-76134200 on Build GRCh38
rs947910324 Chr.15: 76134356-76134356 on Build GRCh38
rs529679850 Chr.15: 76134203-76134203 on Build GRCh38
rs369334168 Chr.15: 76134284-76134284 on Build GRCh38
rs756910465 Chr.15: 76134218-76134218 on Build GRCh38
rs768613347 Chr.15: 76134276-76134276 on Build GRCh38
rs541710365 Chr.15: 76134191-76134191 on Build GRCh38
rs932491629 Chr.15: 76134261-76134261 on Build GRCh38
rs753891578 Chr.15: 76134340-76134340 on Build GRCh38
rs898265846 Chr.15: 76134370-76134370 on Build GRCh38
rs974839536 Chr.15: 76134230-76134230 on Build GRCh38
rs114291110 Chr.15: 76134280-76134280 on Build GRCh38
rs775171421 Chr.15: 76134249-76134250 on Build GRCh38
rs1014287822 Chr.15: 76134183-76134183 on Build GRCh38
rs966685119 Chr.15: 76134197-76134197 on Build GRCh38
rs960490884 Chr.15: 76134308-76134308 on Build GRCh38
rs745365360 Chr.15: 76134245-76134245 on Build GRCh38
rs760661459 Chr.15: 76134312-76134312 on Build GRCh38
rs866619469 Chr.15: 76134278-76134278 on Build GRCh38
rs992357972 Chr.15: 76134310-76134310 on Build GRCh38
rs921128492 Chr.15: 76134232-76134232 on Build GRCh38
rs995687626 Chr.15: 76134371-76134371 on Build GRCh38
rs760068065 Chr.15: 76134350-76134350 on Build GRCh38
rs371301929 Chr.15: 76134321-76134321 on Build GRCh38
rs143633619 Chr.15: 76134184-76134184 on Build GRCh38
rs749288406 Chr.15: 76134267-76134267 on Build GRCh38
rs373141692 Chr.15: 76134293-76134293 on Build GRCh38

View Full Product Details