Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 95114621-95114894 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMPR1B
Entrez Gene ID
658
Gene Name
bone morphogenetic protein receptor type 1B
Gene Aliases
ALK-6, ALK6, AMDD, BDA1D, BDA2, CDw293
UniGene
Hs.598475
Gene Chromosome Location
Chr.4: 94757977-95158453 on Build GRCh38
Transcripts
NM_001203.2 NM_001256792.1 NM_001256793.1 NM_001256794.1 XM_011532201.2 XM_017008558.1 XM_017008559.1 XM_017008560.1 XM_017008561.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4126806 c.222A>C p.L74L Chr.4: 95114798-95114798 on Build GRCh38
4537430 c.241G>A p.G81R Chr.4: 95114727-95114727 on Build GRCh38
4539009 c.262G>T p.E88* Chr.4: 95114748-95114748 on Build GRCh38
448356 c.193C>T p.P65S Chr.4: 95114769-95114769 on Build GRCh38
5167725 c.336+1G>T p.? Chr.4: 95114823-95114823 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs904629575 Chr.4: 95114680-95114680 on Build GRCh38
rs763232227 Chr.4: 95114677-95114677 on Build GRCh38
rs572211990 Chr.4: 95114704-95114704 on Build GRCh38
rs770220808 Chr.4: 95114855-95114855 on Build GRCh38
rs751756955 Chr.4: 95114686-95114686 on Build GRCh38
rs960712934 Chr.4: 95114759-95114759 on Build GRCh38
rs201177627 Chr.4: 95114864-95114864 on Build GRCh38
rs781464761 Chr.4: 95114714-95114714 on Build GRCh38
rs780739446 Chr.4: 95114672-95114675 on Build GRCh38
rs754275762 Chr.4: 95114659-95114660 on Build GRCh38
rs552654246 Chr.4: 95114651-95114652 on Build GRCh38
rs376913841 Chr.4: 95114703-95114703 on Build GRCh38
rs759149974 Chr.4: 95114790-95114790 on Build GRCh38
rs894725180 Chr.4: 95114668-95114668 on Build GRCh38
rs764418758 Chr.4: 95114865-95114865 on Build GRCh38
rs1033648877 Chr.4: 95114740-95114740 on Build GRCh38
rs750279805 Chr.4: 95114659-95114674 on Build GRCh38
rs397802569 Chr.4: 95114669-95114670 on Build GRCh38
rs747805386 Chr.4: 95114673-95114674 on Build GRCh38
rs863225041 Chr.4: 95114733-95114733 on Build GRCh38
rs869115051 Chr.4: 95114670-95114671 on Build GRCh38
rs1017431552 Chr.4: 95114716-95114716 on Build GRCh38
rs750385068 Chr.4: 95114717-95114717 on Build GRCh38
rs111561860 Chr.4: 95114657-95114658 on Build GRCh38
rs200702974 Chr.4: 95114805-95114805 on Build GRCh38
rs749067196 Chr.4: 95114741-95114741 on Build GRCh38
rs765575434 Chr.4: 95114673-95114673 on Build GRCh38
rs778604737 Chr.4: 95114774-95114774 on Build GRCh38
rs1043232365 Chr.4: 95114679-95114679 on Build GRCh38
rs775893590 Chr.4: 95114860-95114860 on Build GRCh38
rs760193561 Chr.4: 95114849-95114849 on Build GRCh38
rs753049798 Chr.4: 95114674-95114674 on Build GRCh38
rs753752689 Chr.4: 95114842-95114842 on Build GRCh38
rs760099912 Chr.4: 95114670-95114670 on Build GRCh38
rs202045659 Chr.4: 95114682-95114682 on Build GRCh38
rs147320212 Chr.4: 95114726-95114726 on Build GRCh38
rs772766219 Chr.4: 95114816-95114816 on Build GRCh38
rs747796220 Chr.4: 95114801-95114801 on Build GRCh38
rs756055104 Chr.4: 95114723-95114723 on Build GRCh38

View Full Product Details