Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 13890274-13890515 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C19orf57
Entrez Gene ID
79173
Gene Name
chromosome 19 open reading frame 57
Gene Aliases
-
UniGene
Hs.143288
Gene Chromosome Location
Chr.19: 13882348-13906125 on Build GRCh38
Transcripts
NM_024323.3 XM_006722885.3 XM_011528286.2 XM_011528287.2 XM_017027300.1 XM_017027301.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5587339 c.571C>T p.P191S Chr.19: 13890285-13890285 on Build GRCh38
1238354 c.477G>A p.T159T Chr.19: 13890379-13890379 on Build GRCh38
119928 c.440T>C p.L147P Chr.19: 13890416-13890416 on Build GRCh38
6008424 c.478G>A p.E160K Chr.19: 13890378-13890378 on Build GRCh38
4501112 c.580C>T p.P194S Chr.19: 13890276-13890276 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs752475723 Chr.19: 13890321-13890321 on Build GRCh38
rs757389709 Chr.19: 13890344-13890344 on Build GRCh38
rs750394425 Chr.19: 13890491-13890491 on Build GRCh38
rs772507081 Chr.19: 13890443-13890443 on Build GRCh38
rs370371789 Chr.19: 13890379-13890379 on Build GRCh38
rs765847377 Chr.19: 13890367-13890367 on Build GRCh38
rs112207239 Chr.19: 13890352-13890352 on Build GRCh38
rs756153411 Chr.19: 13890421-13890421 on Build GRCh38
rs775885327 Chr.19: 13890298-13890298 on Build GRCh38
rs142351146 Chr.19: 13890388-13890388 on Build GRCh38
rs749600312 Chr.19: 13890356-13890356 on Build GRCh38
rs764604383 Chr.19: 13890304-13890304 on Build GRCh38
rs775589300 Chr.19: 13890353-13890353 on Build GRCh38
rs201411314 Chr.19: 13890362-13890362 on Build GRCh38
rs201758211 Chr.19: 13890452-13890452 on Build GRCh38
rs769079703 Chr.19: 13890357-13890357 on Build GRCh38
rs745904967 Chr.19: 13890419-13890419 on Build GRCh38
rs921622831 Chr.19: 13890465-13890465 on Build GRCh38
rs144556741 Chr.19: 13890345-13890345 on Build GRCh38
rs375211344 Chr.19: 13890442-13890442 on Build GRCh38
rs774741315 Chr.19: 13890441-13890441 on Build GRCh38
rs868556045 Chr.19: 13890360-13890366 on Build GRCh38
rs768989799 Chr.19: 13890432-13890432 on Build GRCh38
rs753470005 Chr.19: 13890312-13890312 on Build GRCh38
rs774829475 Chr.19: 13890361-13890361 on Build GRCh38
rs765935675 Chr.19: 13890311-13890311 on Build GRCh38
rs759078366 Chr.19: 13890447-13890447 on Build GRCh38
rs199703310 Chr.19: 13890492-13890492 on Build GRCh38
rs757218856 Chr.19: 13890412-13890412 on Build GRCh38
rs747000381 Chr.19: 13890340-13890340 on Build GRCh38
rs200242396 Chr.19: 13890428-13890428 on Build GRCh38
rs752198106 Chr.19: 13890326-13890327 on Build GRCh38
rs752065500 Chr.19: 13890307-13890307 on Build GRCh38
rs773594583 Chr.19: 13890445-13890445 on Build GRCh38
rs371277207 Chr.19: 13890319-13890319 on Build GRCh38
rs533574843 Chr.19: 13890458-13890458 on Build GRCh38
rs776068415 Chr.19: 13890371-13890371 on Build GRCh38
rs531463341 Chr.19: 13890411-13890411 on Build GRCh38
rs745967261 Chr.19: 13890350-13890350 on Build GRCh38
rs760177608 Chr.19: 13890310-13890310 on Build GRCh38
rs969484545 Chr.19: 13890427-13890427 on Build GRCh38
rs144502446 Chr.19: 13890393-13890393 on Build GRCh38
rs767496887 Chr.19: 13890489-13890489 on Build GRCh38
rs764874450 Chr.19: 13890380-13890380 on Build GRCh38
rs1022357095 Chr.19: 13890305-13890305 on Build GRCh38
rs764766078 Chr.19: 13890309-13890311 on Build GRCh38
rs754702885 Chr.19: 13890314-13890314 on Build GRCh38
rs770020516 Chr.19: 13890296-13890296 on Build GRCh38
rs763250600 Chr.19: 13890303-13890303 on Build GRCh38
rs758128875 Chr.19: 13890334-13890334 on Build GRCh38
rs371641806 Chr.19: 13890454-13890454 on Build GRCh38
rs114174140 Chr.19: 13890415-13890415 on Build GRCh38
rs761715891 Chr.19: 13890488-13890488 on Build GRCh38
rs563035616 Chr.19: 13890453-13890453 on Build GRCh38
rs200387098 Chr.19: 13890468-13890468 on Build GRCh38
rs980157651 Chr.19: 13890376-13890376 on Build GRCh38
rs780319121 Chr.19: 13890426-13890426 on Build GRCh38

View Full Product Details