Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 235441703-235441970 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
B3GALNT2
Entrez Gene ID
148789
Gene Name
beta-1,3-N-acetylgalactosaminyltransferase 2
Gene Aliases
B3GalNAc-T2, MDDGA11
UniGene
Hs.498143
Gene Chromosome Location
Chr.1: 235440654-235504481 on Build GRCh38
Transcripts
XM_006711749.3 NM_001079515.2 NM_001287801.1 NM_001287802.1 NM_003193.4

Gene Symbol
TBCE
Entrez Gene ID
6905
Gene Name
tubulin folding cofactor E
Gene Aliases
HRD, KCS, KCS1, pac2
UniGene
Hs.744998
Gene Chromosome Location
Chr.1: 235367360-235448968 on Build GRCh38
Transcripts
XM_006711749.3 NM_001079515.2 NM_001287801.1 NM_001287802.1 NM_003193.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6797703 c.1277G>C p.G426A Chr.1: 235441820-235441820 on Build GRCh38
3485538 c.1277G>A p.G426D Chr.1: 235441820-235441820 on Build GRCh38
4590579 c.1427A>G p.Y476C Chr.1: 235441817-235441817 on Build GRCh38
4732445 c.1324delA p.N443fs*4 Chr.1: 235441867-235441867 on Build GRCh38
6062166 c.1432G>T p.A478S Chr.1: 235441822-235441822 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs992232386 Chr.1: 235441725-235441725 on Build GRCh38
rs199612067 Chr.1: 235441737-235441740 on Build GRCh38
rs772034880 Chr.1: 235441796-235441797 on Build GRCh38
rs762702207 Chr.1: 235441926-235441926 on Build GRCh38
rs987316259 Chr.1: 235441798-235441798 on Build GRCh38
rs869080776 Chr.1: 235441752-235441755 on Build GRCh38
rs562932013 Chr.1: 235441727-235441727 on Build GRCh38
rs773176639 Chr.1: 235441921-235441921 on Build GRCh38
rs571516848 Chr.1: 235441909-235441909 on Build GRCh38
rs530364039 Chr.1: 235441728-235441728 on Build GRCh38
rs770085515 Chr.1: 235441808-235441808 on Build GRCh38
rs200990578 Chr.1: 235441774-235441774 on Build GRCh38
rs763750658 Chr.1: 235441892-235441897 on Build GRCh38
rs1005936051 Chr.1: 235441795-235441795 on Build GRCh38
rs760144341 Chr.1: 235441763-235441763 on Build GRCh38
rs549442591 Chr.1: 235441939-235441942 on Build GRCh38
rs546638136 Chr.1: 235441908-235441908 on Build GRCh38
rs776558799 Chr.1: 235441791-235441791 on Build GRCh38
rs746518207 Chr.1: 235441892-235441892 on Build GRCh38
rs975889446 Chr.1: 235441906-235441906 on Build GRCh38
rs147789205 Chr.1: 235441724-235441724 on Build GRCh38
rs760204612 Chr.1: 235441876-235441878 on Build GRCh38
rs373701959 Chr.1: 235441768-235441768 on Build GRCh38
rs935204491 Chr.1: 235441744-235441744 on Build GRCh38
rs768119813 Chr.1: 235441903-235441903 on Build GRCh38
rs960774634 Chr.1: 235441722-235441722 on Build GRCh38
rs754330018 Chr.1: 235441889-235441889 on Build GRCh38
rs753778541 Chr.1: 235441765-235441765 on Build GRCh38
rs910364188 Chr.1: 235441799-235441799 on Build GRCh38
rs776299404 Chr.1: 235441925-235441925 on Build GRCh38
rs764071956 Chr.1: 235441764-235441764 on Build GRCh38
rs964186057 Chr.1: 235441806-235441806 on Build GRCh38
rs765079996 Chr.1: 235441917-235441917 on Build GRCh38
rs185752881 Chr.1: 235441757-235441757 on Build GRCh38
rs771725945 Chr.1: 235441863-235441863 on Build GRCh38
rs778133382 Chr.1: 235441862-235441862 on Build GRCh38
rs775360097 Chr.1: 235441842-235441848 on Build GRCh38
rs761697580 Chr.1: 235441767-235441767 on Build GRCh38
rs79490393 Chr.1: 235441776-235441776 on Build GRCh38
rs745665486 Chr.1: 235441780-235441781 on Build GRCh38
rs925148317 Chr.1: 235441737-235441744 on Build GRCh38

View Full Product Details