Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 123724603-123724866 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP6V0A2
Entrez Gene ID
23545
Gene Name
ATPase H+ transporting V0 subunit a2
Gene Aliases
A2, ARCL, ARCL2A, ATP6A2, ATP6N1D, J6B7, RTF, STV1, TJ6, TJ6M, TJ6S, VPH1, WSS
UniGene
Hs.25786
Gene Chromosome Location
Chr.12: 123712318-123761755 on Build GRCh38
Transcripts
NM_012463.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
692501 c.381G>T p.E127D Chr.12: 123724740-123724740 on Build GRCh38
2226066 c.309G>T p.K103N Chr.12: 123724668-123724668 on Build GRCh38
1359774 c.328G>T p.E110* Chr.12: 123724687-123724687 on Build GRCh38
6608041 c.394C>G p.L132V Chr.12: 123724753-123724753 on Build GRCh38
936482 c.422G>A p.R141H Chr.12: 123724781-123724781 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs377601811 Chr.12: 123724675-123724675 on Build GRCh38
rs1139789 Chr.12: 123724785-123724785 on Build GRCh38
rs768807230 Chr.12: 123724784-123724784 on Build GRCh38
rs752200097 Chr.12: 123724744-123724744 on Build GRCh38
rs202106256 Chr.12: 123724765-123724765 on Build GRCh38
rs140223278 Chr.12: 123724732-123724732 on Build GRCh38
rs777386682 Chr.12: 123724749-123724749 on Build GRCh38
rs778329530 Chr.12: 123724639-123724639 on Build GRCh38
rs925809255 Chr.12: 123724721-123724721 on Build GRCh38
rs774904501 Chr.12: 123724711-123724711 on Build GRCh38
rs751125314 Chr.12: 123724757-123724757 on Build GRCh38
rs752508701 Chr.12: 123724828-123724835 on Build GRCh38
rs752361719 Chr.12: 123724641-123724641 on Build GRCh38
rs763329850 Chr.12: 123724692-123724692 on Build GRCh38
rs563333869 Chr.12: 123724671-123724671 on Build GRCh38
rs199801221 Chr.12: 123724747-123724747 on Build GRCh38
rs373770878 Chr.12: 123724651-123724651 on Build GRCh38
rs11837144 Chr.12: 123724805-123724805 on Build GRCh38
rs773435544 Chr.12: 123724800-123724800 on Build GRCh38
rs936214622 Chr.12: 123724786-123724786 on Build GRCh38
rs769845216 Chr.12: 123724780-123724780 on Build GRCh38
rs794727643 Chr.12: 123724663-123724663 on Build GRCh38
rs535259667 Chr.12: 123724803-123724803 on Build GRCh38
rs780788821 Chr.12: 123724769-123724769 on Build GRCh38
rs777613164 Chr.12: 123724656-123724656 on Build GRCh38
rs763239634 Chr.12: 123724677-123724678 on Build GRCh38
rs745872717 Chr.12: 123724770-123724770 on Build GRCh38
rs772500257 Chr.12: 123724795-123724795 on Build GRCh38
rs773252881 Chr.12: 123724643-123724643 on Build GRCh38
rs767732870 Chr.12: 123724730-123724730 on Build GRCh38
rs1047744059 Chr.12: 123724764-123724764 on Build GRCh38
rs761873132 Chr.12: 123724789-123724789 on Build GRCh38
rs767257316 Chr.12: 123724747-123724748 on Build GRCh38
rs551494374 Chr.12: 123724657-123724657 on Build GRCh38
rs80356752 Chr.12: 123724712-123724713 on Build GRCh38
rs764787363 Chr.12: 123724741-123724741 on Build GRCh38
rs899705414 Chr.12: 123724797-123724797 on Build GRCh38
rs754545836 Chr.12: 123724635-123724635 on Build GRCh38
rs143509747 Chr.12: 123724781-123724781 on Build GRCh38
rs762475349 Chr.12: 123724806-123724806 on Build GRCh38
rs144499089 Chr.12: 123724668-123724668 on Build GRCh38
rs937389279 Chr.12: 123724734-123724734 on Build GRCh38
rs769927427 Chr.12: 123724672-123724672 on Build GRCh38
rs374768446 Chr.12: 123724833-123724833 on Build GRCh38
rs762260351 Chr.12: 123724727-123724727 on Build GRCh38
rs761030937 Chr.12: 123724740-123724740 on Build GRCh38

View Full Product Details