Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 107755517-107755766 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BBX
Entrez Gene ID
56987
Gene Name
BBX, HMG-box containing
Gene Aliases
ARTC1, HBP2, HSPC339, MDS001
UniGene
Hs.124366
Gene Chromosome Location
Chr.3: 107522936-107811329 on Build GRCh38
Transcripts
NM_001142568.2 NM_001276286.1 NM_020235.6 XM_005247642.3 XM_005247643.3 XM_005247644.3 XM_011512999.2 XM_011513000.1 XM_011513001.1 XM_011513002.2 XM_011513003.2 XM_011513004.2 XM_011513005.2 XM_011513006.2 XM_011513007.2 XM_011513010.2 XM_017006881.1 XM_017006882.1 XM_017006883.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4838011 c.833C>G p.S278* Chr.3: 107755605-107755605 on Build GRCh38
6162808 c.886G>T p.A296S Chr.3: 107755658-107755658 on Build GRCh38
2916572 c.857C>T p.A286V Chr.3: 107755629-107755629 on Build GRCh38
4838010 c.833C>G p.S278* Chr.3: 107755605-107755605 on Build GRCh38
3800851 c.895C>T p.Q299* Chr.3: 107755667-107755667 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs369165652 Chr.3: 107755603-107755603 on Build GRCh38
rs745535182 Chr.3: 107755593-107755594 on Build GRCh38
rs771796024 Chr.3: 107755647-107755647 on Build GRCh38
rs771034767 Chr.3: 107755722-107755722 on Build GRCh38
rs756408349 Chr.3: 107755714-107755714 on Build GRCh38
rs754988118 Chr.3: 107755570-107755570 on Build GRCh38
rs111300616 Chr.3: 107755552-107755552 on Build GRCh38
rs749737625 Chr.3: 107755618-107755618 on Build GRCh38
rs780232853 Chr.3: 107755718-107755718 on Build GRCh38
rs373312172 Chr.3: 107755687-107755687 on Build GRCh38
rs761415898 Chr.3: 107755643-107755643 on Build GRCh38
rs538786093 Chr.3: 107755699-107755699 on Build GRCh38
rs951202577 Chr.3: 107755644-107755644 on Build GRCh38
rs757046096 Chr.3: 107755610-107755610 on Build GRCh38
rs899559762 Chr.3: 107755593-107755593 on Build GRCh38
rs747121201 Chr.3: 107755585-107755585 on Build GRCh38
rs750613004 Chr.3: 107755712-107755712 on Build GRCh38
rs778368925 Chr.3: 107755553-107755553 on Build GRCh38
rs773696884 Chr.3: 107755628-107755628 on Build GRCh38
rs760475004 Chr.3: 107755658-107755658 on Build GRCh38
rs757314003 Chr.3: 107755697-107755697 on Build GRCh38
rs769229464 Chr.3: 107755619-107755619 on Build GRCh38
rs757401820 Chr.3: 107755598-107755598 on Build GRCh38
rs373232502 Chr.3: 107755615-107755615 on Build GRCh38
rs766061685 Chr.3: 107755669-107755669 on Build GRCh38
rs868711354 Chr.3: 107755609-107755609 on Build GRCh38
rs772990530 Chr.3: 107755653-107755653 on Build GRCh38
rs376143920 Chr.3: 107755679-107755679 on Build GRCh38
rs749695398 Chr.3: 107755720-107755720 on Build GRCh38
rs770138977 Chr.3: 107755614-107755614 on Build GRCh38
rs751578589 Chr.3: 107755695-107755695 on Build GRCh38
rs78247457 Chr.3: 107755700-107755700 on Build GRCh38
rs753685858 Chr.3: 107755670-107755670 on Build GRCh38
rs138605547 Chr.3: 107755607-107755607 on Build GRCh38
rs148091529 Chr.3: 107755739-107755742 on Build GRCh38
rs370340696 Chr.3: 107755709-107755709 on Build GRCh38
rs777662857 Chr.3: 107755576-107755576 on Build GRCh38

View Full Product Details