Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 2369265-2369536 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EIF3B
Entrez Gene ID
8662
Gene Name
eukaryotic translation initiation factor 3 subunit B
Gene Aliases
EIF3-ETA, EIF3-P110, EIF3-P116, EIF3S9, PRT1
UniGene
Hs.371001
Gene Chromosome Location
Chr.7: 2354540-2380745 on Build GRCh38
Transcripts
NM_001037283.1 NM_003751.3 XM_011515599.1 XM_011515600.1 XM_011515601.1 XM_017012752.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6652796 c.1408T>C p.F470L Chr.7: 2369476-2369476 on Build GRCh38
1698324 c.1414T>C p.W472R Chr.7: 2369482-2369482 on Build GRCh38
5407259 c.1420C>T p.P474S Chr.7: 2369488-2369488 on Build GRCh38
3637572 c.1412C>T p.S471F Chr.7: 2369480-2369480 on Build GRCh38
6836973 c.1445G>T p.W482L Chr.7: 2369513-2369513 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753205309 Chr.7: 2369402-2369402 on Build GRCh38
rs1004213958 Chr.7: 2369349-2369349 on Build GRCh38
rs186633772 Chr.7: 2369351-2369351 on Build GRCh38
rs200537132 Chr.7: 2369433-2369433 on Build GRCh38
rs60545087 Chr.7: 2369468-2369468 on Build GRCh38
rs907846912 Chr.7: 2369423-2369423 on Build GRCh38
rs201839393 Chr.7: 2369481-2369481 on Build GRCh38
rs142311273 Chr.7: 2369496-2369496 on Build GRCh38
rs191086790 Chr.7: 2369461-2369461 on Build GRCh38
rs948914293 Chr.7: 2369289-2369289 on Build GRCh38
rs772377420 Chr.7: 2369376-2369378 on Build GRCh38
rs557838273 Chr.7: 2369382-2369382 on Build GRCh38
rs761204844 Chr.7: 2369432-2369432 on Build GRCh38
rs2301944 Chr.7: 2369400-2369400 on Build GRCh38
rs200382824 Chr.7: 2369368-2369368 on Build GRCh38
rs146706009 Chr.7: 2369313-2369313 on Build GRCh38
rs766507364 Chr.7: 2369331-2369331 on Build GRCh38
rs930913349 Chr.7: 2369312-2369312 on Build GRCh38
rs530514721 Chr.7: 2369311-2369311 on Build GRCh38
rs1054273438 Chr.7: 2369295-2369295 on Build GRCh38
rs549064666 Chr.7: 2369361-2369361 on Build GRCh38
rs774094818 Chr.7: 2369490-2369490 on Build GRCh38
rs749142613 Chr.7: 2369469-2369469 on Build GRCh38
rs540619191 Chr.7: 2369397-2369397 on Build GRCh38
rs758534447 Chr.7: 2369462-2369462 on Build GRCh38
rs773450314 Chr.7: 2369422-2369423 on Build GRCh38
rs753913331 Chr.7: 2369371-2369371 on Build GRCh38
rs755283834 Chr.7: 2369443-2369443 on Build GRCh38
rs991851821 Chr.7: 2369392-2369392 on Build GRCh38
rs564781690 Chr.7: 2369314-2369314 on Build GRCh38
rs772779770 Chr.7: 2369506-2369506 on Build GRCh38
rs1040503187 Chr.7: 2369354-2369354 on Build GRCh38
rs146119314 Chr.7: 2369505-2369505 on Build GRCh38
rs754320382 Chr.7: 2369434-2369434 on Build GRCh38
rs903731676 Chr.7: 2369377-2369377 on Build GRCh38
rs984278698 Chr.7: 2369373-2369373 on Build GRCh38
rs917402747 Chr.7: 2369286-2369286 on Build GRCh38
rs115449632 Chr.7: 2369355-2369355 on Build GRCh38
rs112580498 Chr.7: 2369379-2369379 on Build GRCh38
rs768886455 Chr.7: 2369438-2369442 on Build GRCh38
rs915743522 Chr.7: 2369460-2369460 on Build GRCh38
rs368880701 Chr.7: 2369456-2369456 on Build GRCh38
rs758725837 Chr.7: 2369413-2369413 on Build GRCh38
rs773888724 Chr.7: 2369426-2369426 on Build GRCh38
rs150290110 Chr.7: 2369425-2369425 on Build GRCh38
rs547187102 Chr.7: 2369348-2369348 on Build GRCh38
rs183020610 Chr.7: 2369321-2369321 on Build GRCh38
rs1044554719 Chr.7: 2369302-2369302 on Build GRCh38
rs768314023 Chr.7: 2369480-2369480 on Build GRCh38
rs538216606 Chr.7: 2369381-2369381 on Build GRCh38
rs777778913 Chr.7: 2369466-2369466 on Build GRCh38
rs532553945 Chr.7: 2369424-2369424 on Build GRCh38
rs983746269 Chr.7: 2369414-2369414 on Build GRCh38
rs920811620 Chr.7: 2369412-2369412 on Build GRCh38

View Full Product Details