Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 27344924-27345159 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL4R
Entrez Gene ID
3566
Gene Name
interleukin 4 receptor
Gene Aliases
CD124, IL-4RA, IL4RA
UniGene
Hs.513457
Gene Chromosome Location
Chr.16: 27313668-27364778 on Build GRCh38
Transcripts
NM_000418.3 NM_001257406.1 NM_001257407.1 NM_001257997.1 XM_011545825.1 XM_011545826.2 XM_011545827.2 XM_011545828.2 XM_011545829.1 XM_011545830.2 XM_011545831.1 XM_011545832.1 XM_011545833.2 XM_011545834.2 XM_017023211.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7127136 c.337G>T p.G113C Chr.16: 27344996-27344996 on Build GRCh38
6397015 c.317G>A p.G106E Chr.16: 27344976-27344976 on Build GRCh38
6685414 c.277G>A p.V93M Chr.16: 27344936-27344936 on Build GRCh38
3108721 c.341C>T p.S114F Chr.16: 27345000-27345000 on Build GRCh38
4489293 c.345C>T p.F115F Chr.16: 27345004-27345004 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs771597376 Chr.16: 27345042-27345042 on Build GRCh38
rs746593366 Chr.16: 27345032-27345032 on Build GRCh38
rs761781246 Chr.16: 27344952-27344952 on Build GRCh38
rs574446159 Chr.16: 27344998-27344998 on Build GRCh38
rs766157629 Chr.16: 27345061-27345061 on Build GRCh38
rs762694127 Chr.16: 27344993-27344993 on Build GRCh38
rs543167942 Chr.16: 27345000-27345000 on Build GRCh38
rs888973268 Chr.16: 27345047-27345047 on Build GRCh38
rs960324104 Chr.16: 27345081-27345081 on Build GRCh38
rs17548711 Chr.16: 27345093-27345093 on Build GRCh38
rs750189532 Chr.16: 27344960-27344960 on Build GRCh38
rs779751751 Chr.16: 27345031-27345031 on Build GRCh38
rs1008636293 Chr.16: 27345055-27345055 on Build GRCh38
rs897471715 Chr.16: 27345089-27345089 on Build GRCh38
rs776228073 Chr.16: 27345039-27345039 on Build GRCh38
rs753929235 Chr.16: 27345010-27345010 on Build GRCh38
rs576739360 Chr.16: 27345012-27345012 on Build GRCh38
rs77491333 Chr.16: 27345043-27345043 on Build GRCh38
rs779320253 Chr.16: 27345013-27345013 on Build GRCh38
rs1012128011 Chr.16: 27345082-27345082 on Build GRCh38
rs1050629895 Chr.16: 27344986-27344986 on Build GRCh38
rs112497527 Chr.16: 27344950-27344950 on Build GRCh38
rs1034710194 Chr.16: 27345053-27345053 on Build GRCh38
rs2072130 Chr.16: 27345077-27345077 on Build GRCh38
rs369875568 Chr.16: 27345044-27345044 on Build GRCh38
rs2074571 Chr.16: 27345091-27345091 on Build GRCh38
rs991472366 Chr.16: 27344999-27344999 on Build GRCh38
rs773075015 Chr.16: 27345048-27345048 on Build GRCh38
rs1011518566 Chr.16: 27345092-27345092 on Build GRCh38
rs141903283 Chr.16: 27345014-27345014 on Build GRCh38
rs760218604 Chr.16: 27344946-27344946 on Build GRCh38
rs199703000 Chr.16: 27345002-27345002 on Build GRCh38
rs570293541 Chr.16: 27345133-27345133 on Build GRCh38
rs2074572 Chr.16: 27345038-27345038 on Build GRCh38
rs565620025 Chr.16: 27345070-27345070 on Build GRCh38
rs1049657568 Chr.16: 27345127-27345127 on Build GRCh38
rs375589999 Chr.16: 27345112-27345112 on Build GRCh38
rs758746097 Chr.16: 27345019-27345019 on Build GRCh38
rs765284416 Chr.16: 27344959-27344959 on Build GRCh38
rs55832501 Chr.16: 27345049-27345049 on Build GRCh38

View Full Product Details