Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 158736979-158737242 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ESYT2
Entrez Gene ID
57488
Gene Name
extended synaptotagmin 2
Gene Aliases
CHR2SYT, E-Syt2, FAM62B
UniGene
Hs.490795
Gene Chromosome Location
Chr.7: 158730998-158829628 on Build GRCh38
Transcripts
NM_020728.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6109403 c.2473G>T p.D825Y Chr.7: 158737055-158737055 on Build GRCh38
452800 c.2427A>C p.G809G Chr.7: 158737101-158737101 on Build GRCh38
4872997 c.724C>T p.R242W Chr.7: 158737109-158737109 on Build GRCh38
4481016 c.2476C>T p.Q826* Chr.7: 158737052-158737052 on Build GRCh38
5345312 c.2436A>C p.K812N Chr.7: 158737092-158737092 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs985521809 Chr.7: 158737093-158737093 on Build GRCh38
rs148294938 Chr.7: 158737041-158737041 on Build GRCh38
rs574320442 Chr.7: 158737027-158737027 on Build GRCh38
rs774403218 Chr.7: 158737146-158737146 on Build GRCh38
rs779540208 Chr.7: 158737010-158737010 on Build GRCh38
rs755359602 Chr.7: 158737085-158737085 on Build GRCh38
rs371348949 Chr.7: 158737043-158737043 on Build GRCh38
rs143047782 Chr.7: 158737131-158737131 on Build GRCh38
rs768471495 Chr.7: 158737136-158737136 on Build GRCh38
rs761451501 Chr.7: 158737047-158737047 on Build GRCh38
rs747048998 Chr.7: 158737099-158737099 on Build GRCh38
rs972122996 Chr.7: 158737217-158737217 on Build GRCh38
rs377389303 Chr.7: 158737209-158737209 on Build GRCh38
rs749021409 Chr.7: 158737216-158737216 on Build GRCh38
rs142637211 Chr.7: 158737096-158737096 on Build GRCh38
rs375153914 Chr.7: 158737044-158737044 on Build GRCh38
rs369510097 Chr.7: 158737125-158737125 on Build GRCh38
rs369892502 Chr.7: 158737218-158737218 on Build GRCh38
rs528398874 Chr.7: 158737175-158737175 on Build GRCh38
rs761206385 Chr.7: 158737151-158737151 on Build GRCh38
rs746050325 Chr.7: 158737123-158737123 on Build GRCh38
rs756653279 Chr.7: 158737042-158737042 on Build GRCh38
rs749647863 Chr.7: 158737182-158737183 on Build GRCh38
rs755451473 Chr.7: 158737005-158737005 on Build GRCh38
rs776900417 Chr.7: 158737024-158737024 on Build GRCh38
rs894773771 Chr.7: 158737184-158737184 on Build GRCh38
rs757736356 Chr.7: 158737194-158737194 on Build GRCh38
rs2305467 Chr.7: 158737198-158737198 on Build GRCh38
rs749115249 Chr.7: 158737135-158737135 on Build GRCh38
rs775209416 Chr.7: 158737133-158737133 on Build GRCh38
rs780628638 Chr.7: 158737047-158737047 on Build GRCh38
rs762751557 Chr.7: 158737042-158737042 on Build GRCh38
rs766140607 Chr.7: 158737164-158737164 on Build GRCh38
rs756264220 Chr.7: 158737203-158737203 on Build GRCh38
rs368810393 Chr.7: 158737018-158737018 on Build GRCh38
rs987437502 Chr.7: 158737190-158737190 on Build GRCh38
rs765653857 Chr.7: 158737086-158737086 on Build GRCh38
rs781200960 Chr.7: 158737197-158737197 on Build GRCh38
rs931986940 Chr.7: 158737012-158737012 on Build GRCh38
rs147587512 Chr.7: 158737155-158737155 on Build GRCh38
rs147062200 Chr.7: 158737161-158737161 on Build GRCh38
rs753130973 Chr.7: 158737089-158737089 on Build GRCh38
rs752152653 Chr.7: 158737189-158737189 on Build GRCh38
rs772661672 Chr.7: 158737014-158737014 on Build GRCh38
rs757411199 Chr.7: 158737107-158737107 on Build GRCh38
rs750149909 Chr.7: 158737083-158737083 on Build GRCh38
rs6960873 Chr.7: 158737013-158737013 on Build GRCh38
rs769618054 Chr.7: 158737030-158737030 on Build GRCh38
rs899412260 Chr.7: 158737141-158737141 on Build GRCh38
rs753041016 Chr.7: 158737171-158737171 on Build GRCh38
rs771057217 Chr.7: 158737020-158737020 on Build GRCh38
rs556761451 Chr.7: 158737108-158737108 on Build GRCh38
rs758770896 Chr.7: 158737094-158737094 on Build GRCh38
rs777662766 Chr.7: 158737015-158737015 on Build GRCh38
rs1009955193 Chr.7: 158737056-158737056 on Build GRCh38
rs767062736 Chr.7: 158737067-158737067 on Build GRCh38
rs764490274 Chr.7: 158737187-158737187 on Build GRCh38

View Full Product Details