Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 125910443-125910716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DDX25
Entrez Gene ID
29118
Gene Name
DEAD-box helicase 25
Gene Aliases
GRTH
UniGene
Hs.420263
Gene Chromosome Location
Chr.11: 125903275-125923110 on Build GRCh38
Transcripts
NM_013264.4 XM_011542790.2 XM_011542791.2 XM_017017626.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
122534 c.247G>C p.V83L Chr.11: 125910445-125910445 on Build GRCh38
1975826 c.610C>T p.R204* Chr.11: 125910466-125910466 on Build GRCh38
5790861 c.249T>C p.V83V Chr.11: 125910447-125910447 on Build GRCh38
5986970 c.272G>A p.G91E Chr.11: 125910470-125910470 on Build GRCh38
5790860 c.591T>C p.V197V Chr.11: 125910447-125910447 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs935774889 Chr.11: 125910479-125910479 on Build GRCh38
rs1041241134 Chr.11: 125910669-125910669 on Build GRCh38
rs143200532 Chr.11: 125910628-125910628 on Build GRCh38
rs758095951 Chr.11: 125910487-125910487 on Build GRCh38
rs776047420 Chr.11: 125910477-125910477 on Build GRCh38
rs1012514310 Chr.11: 125910565-125910565 on Build GRCh38
rs370940701 Chr.11: 125910476-125910476 on Build GRCh38
rs753116733 Chr.11: 125910469-125910469 on Build GRCh38
rs1023845322 Chr.11: 125910618-125910618 on Build GRCh38
rs575001091 Chr.11: 125910639-125910639 on Build GRCh38
rs1054199923 Chr.11: 125910526-125910526 on Build GRCh38
rs1015530362 Chr.11: 125910644-125910644 on Build GRCh38
rs769949571 Chr.11: 125910511-125910511 on Build GRCh38
rs537108516 Chr.11: 125910643-125910643 on Build GRCh38
rs962222076 Chr.11: 125910655-125910655 on Build GRCh38
rs756842297 Chr.11: 125910480-125910480 on Build GRCh38
rs920481123 Chr.11: 125910531-125910531 on Build GRCh38
rs755702115 Chr.11: 125910475-125910475 on Build GRCh38
rs376323512 Chr.11: 125910482-125910482 on Build GRCh38
rs1027715720 Chr.11: 125910687-125910687 on Build GRCh38
rs973908741 Chr.11: 125910659-125910659 on Build GRCh38
rs894159930 Chr.11: 125910532-125910532 on Build GRCh38
rs768141649 Chr.11: 125910468-125910468 on Build GRCh38
rs760391003 Chr.11: 125910538-125910538 on Build GRCh38
rs746871138 Chr.11: 125910498-125910498 on Build GRCh38
rs80214587 Chr.11: 125910642-125910642 on Build GRCh38
rs780088137 Chr.11: 125910492-125910492 on Build GRCh38

View Full Product Details