Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 80181456-80181715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CARD14
Entrez Gene ID
79092
Gene Name
caspase recruitment domain family member 14
Gene Aliases
BIMP2, CARMA2, PRP, PSORS2, PSS1
UniGene
Hs.675480
Gene Chromosome Location
Chr.17: 80169992-80209331 on Build GRCh38
Transcripts
NM_001257970.1 NM_024110.4 NR_047566.1 XM_011525212.1 XM_011525213.1 XM_011525215.1 XM_011525216.1 XM_011525217.1 XM_011525218.2 XM_017025057.1 XM_017025058.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6283054 c.95G>A p.R32H Chr.17: 80181533-80181533 on Build GRCh38
4884496 c.156G>T p.L52L Chr.17: 80181594-80181594 on Build GRCh38
3787464 c.45C>T p.D15D Chr.17: 80181483-80181483 on Build GRCh38
3787465 c.110G>A p.S37N Chr.17: 80181548-80181548 on Build GRCh38
6620116 c.31C>A p.L11I Chr.17: 80181469-80181469 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755140319 Chr.17: 80181598-80181598 on Build GRCh38
rs539713605 Chr.17: 80181669-80181669 on Build GRCh38
rs374690955 Chr.17: 80181568-80181568 on Build GRCh38
rs1043628741 Chr.17: 80181640-80181640 on Build GRCh38
rs549347508 Chr.17: 80181528-80181528 on Build GRCh38
rs776565097 Chr.17: 80181491-80181491 on Build GRCh38
rs747222317 Chr.17: 80181474-80181474 on Build GRCh38
rs281875217 Chr.17: 80181550-80181550 on Build GRCh38
rs770895364 Chr.17: 80181656-80181656 on Build GRCh38
rs748158215 Chr.17: 80181634-80181634 on Build GRCh38
rs746969281 Chr.17: 80181569-80181569 on Build GRCh38
rs1039798712 Chr.17: 80181619-80181619 on Build GRCh38
rs553686312 Chr.17: 80181622-80181622 on Build GRCh38
rs1043700167 Chr.17: 80181507-80181507 on Build GRCh38
rs368565321 Chr.17: 80181597-80181597 on Build GRCh38
rs768959428 Chr.17: 80181483-80181483 on Build GRCh38
rs934542227 Chr.17: 80181524-80181524 on Build GRCh38
rs752137635 Chr.17: 80181555-80181555 on Build GRCh38
rs777880019 Chr.17: 80181552-80181552 on Build GRCh38
rs895516485 Chr.17: 80181649-80181649 on Build GRCh38
rs895639722 Chr.17: 80181532-80181532 on Build GRCh38
rs963268943 Chr.17: 80181518-80181518 on Build GRCh38
rs1014127464 Chr.17: 80181543-80181543 on Build GRCh38
rs145933018 Chr.17: 80181588-80181588 on Build GRCh38
rs868109242 Chr.17: 80181546-80181546 on Build GRCh38
rs537669777 Chr.17: 80181494-80181494 on Build GRCh38
rs371643272 Chr.17: 80181551-80181551 on Build GRCh38
rs868628070 Chr.17: 80181583-80181583 on Build GRCh38
rs983653572 Chr.17: 80181618-80181618 on Build GRCh38
rs369726105 Chr.17: 80181644-80181644 on Build GRCh38
rs115582620 Chr.17: 80181623-80181623 on Build GRCh38
rs775216045 Chr.17: 80181690-80181690 on Build GRCh38
rs1012573570 Chr.17: 80181655-80181655 on Build GRCh38
rs772077307 Chr.17: 80181529-80181529 on Build GRCh38
rs773633754 Chr.17: 80181641-80181641 on Build GRCh38
rs773137252 Chr.17: 80181501-80181501 on Build GRCh38
rs770888241 Chr.17: 80181637-80181637 on Build GRCh38
rs375624435 Chr.17: 80181643-80181643 on Build GRCh38
rs577286599 Chr.17: 80181648-80181648 on Build GRCh38
rs569889639 Chr.17: 80181473-80181473 on Build GRCh38
rs759434673 Chr.17: 80181689-80181689 on Build GRCh38
rs1053020506 Chr.17: 80181672-80181672 on Build GRCh38
rs748315074 Chr.17: 80181492-80181492 on Build GRCh38
rs762697011 Chr.17: 80181508-80181508 on Build GRCh38
rs751003264 Chr.17: 80181585-80181585 on Build GRCh38
rs774252063 Chr.17: 80181537-80181537 on Build GRCh38
rs759080738 Chr.17: 80181545-80181545 on Build GRCh38
rs919076101 Chr.17: 80181533-80181533 on Build GRCh38
rs979943482 Chr.17: 80181654-80181654 on Build GRCh38
rs568127402 Chr.17: 80181587-80181587 on Build GRCh38

View Full Product Details