Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 49016627-49016874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DALRD3
Entrez Gene ID
55152
Gene Name
DALR anticodon binding domain containing 3
Gene Aliases
-
UniGene
Hs.654815
Gene Chromosome Location
Chr.3: 49015399-49021386 on Build GRCh38
Transcripts
NM_001009996.2 NM_001276405.1 NM_018114.5 XM_005265269.3 XM_006713219.2 XM_011533891.1 XM_011533892.1 XM_011533893.1 XM_017006723.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1495550 c.533C>T p.A178V Chr.3: 49016641-49016641 on Build GRCh38
6991582 c.1024A>G p.K342E Chr.3: 49016651-49016651 on Build GRCh38
4823549 c.450G>A p.V150V Chr.3: 49016824-49016824 on Build GRCh38
1249671 c.494_500+2delACTACGAGT p.? Chr.3: 49016772-49016780 on Build GRCh38
1249670 c.995_1001+2delACTACGAGT p.? Chr.3: 49016772-49016780 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748259680 Chr.3: 49016733-49016734 on Build GRCh38
rs532179385 Chr.3: 49016715-49016715 on Build GRCh38
rs763357889 Chr.3: 49016681-49016681 on Build GRCh38
rs200954716 Chr.3: 49016767-49016767 on Build GRCh38
rs749624707 Chr.3: 49016842-49016842 on Build GRCh38
rs773642303 Chr.3: 49016827-49016827 on Build GRCh38
rs151003537 Chr.3: 49016784-49016784 on Build GRCh38
rs769922382 Chr.3: 49016764-49016765 on Build GRCh38
rs200110761 Chr.3: 49016800-49016800 on Build GRCh38
rs753509171 Chr.3: 49016779-49016779 on Build GRCh38
rs768129897 Chr.3: 49016818-49016818 on Build GRCh38
rs771733344 Chr.3: 49016753-49016753 on Build GRCh38
rs202179688 Chr.3: 49016669-49016669 on Build GRCh38
rs757198044 Chr.3: 49016785-49016785 on Build GRCh38
rs759250055 Chr.3: 49016660-49016660 on Build GRCh38
rs770474292 Chr.3: 49016658-49016658 on Build GRCh38
rs565800639 Chr.3: 49016850-49016850 on Build GRCh38
rs757953832 Chr.3: 49016802-49016802 on Build GRCh38
rs890657487 Chr.3: 49016821-49016821 on Build GRCh38
rs775167331 Chr.3: 49016754-49016754 on Build GRCh38
rs536329823 Chr.3: 49016854-49016854 on Build GRCh38
rs751626672 Chr.3: 49016690-49016690 on Build GRCh38
rs766678724 Chr.3: 49016853-49016854 on Build GRCh38
rs746619703 Chr.3: 49016756-49016756 on Build GRCh38
rs139901939 Chr.3: 49016775-49016775 on Build GRCh38
rs776511792 Chr.3: 49016723-49016724 on Build GRCh38
rs771601619 Chr.3: 49016843-49016843 on Build GRCh38
rs756273977 Chr.3: 49016776-49016776 on Build GRCh38
rs747151188 Chr.3: 49016705-49016706 on Build GRCh38
rs1008980006 Chr.3: 49016828-49016828 on Build GRCh38
rs371893230 Chr.3: 49016771-49016771 on Build GRCh38
rs778397200 Chr.3: 49016733-49016733 on Build GRCh38
rs971529201 Chr.3: 49016728-49016728 on Build GRCh38
rs745322798 Chr.3: 49016737-49016737 on Build GRCh38
rs764014482 Chr.3: 49016777-49016777 on Build GRCh38
rs766937042 Chr.3: 49016769-49016769 on Build GRCh38
rs767718771 Chr.3: 49016694-49016694 on Build GRCh38
rs750988575 Chr.3: 49016677-49016677 on Build GRCh38
rs550071849 Chr.3: 49016666-49016666 on Build GRCh38
rs770252356 Chr.3: 49016757-49016757 on Build GRCh38
rs759652542 Chr.3: 49016772-49016772 on Build GRCh38
rs763480847 Chr.3: 49016811-49016812 on Build GRCh38
rs760849977 Chr.3: 49016717-49016717 on Build GRCh38
rs757179521 Chr.3: 49016722-49016722 on Build GRCh38
rs139323774 Chr.3: 49016667-49016667 on Build GRCh38
rs746745090 Chr.3: 49016815-49016815 on Build GRCh38
rs778773139 Chr.3: 49016792-49016792 on Build GRCh38
rs368501664 Chr.3: 49016685-49016685 on Build GRCh38
rs763561130 Chr.3: 49016768-49016768 on Build GRCh38
rs755267600 Chr.3: 49016691-49016691 on Build GRCh38
rs765488745 Chr.3: 49016676-49016676 on Build GRCh38
rs749302759 Chr.3: 49016720-49016720 on Build GRCh38
rs779418459 Chr.3: 49016812-49016812 on Build GRCh38
rs773689750 Chr.3: 49016785-49016786 on Build GRCh38
rs775841510 Chr.3: 49016708-49016708 on Build GRCh38
rs768936976 Chr.3: 49016717-49016718 on Build GRCh38
rs749144961 Chr.3: 49016655-49016655 on Build GRCh38

View Full Product Details