Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 155506933-155507202 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CNPY1
Entrez Gene ID
285888
Gene Name
canopy FGF signaling regulator 1
Gene Aliases
-
UniGene
Hs.146751
Gene Chromosome Location
Chr.7: 155501258-155533844 on Build GRCh38
Transcripts
NM_001103176.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6013027 c.232G>A p.E78K Chr.7: 155507029-155507029 on Build GRCh38
116065 c.187C>T p.L63F Chr.7: 155507074-155507074 on Build GRCh38
1449601 c.241+2T>C p.? Chr.7: 155507018-155507018 on Build GRCh38
1087816 c.192C>T p.I64I Chr.7: 155507069-155507069 on Build GRCh38
5872336 c.196delC p.Q66fs*6 Chr.7: 155507065-155507065 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs764781986 Chr.7: 155506990-155506990 on Build GRCh38
rs368748286 Chr.7: 155506985-155506985 on Build GRCh38
rs755298921 Chr.7: 155507019-155507019 on Build GRCh38
rs762626348 Chr.7: 155507104-155507104 on Build GRCh38
rs367713933 Chr.7: 155507152-155507152 on Build GRCh38
rs555672987 Chr.7: 155507070-155507070 on Build GRCh38
rs766758654 Chr.7: 155506972-155506972 on Build GRCh38
rs779011022 Chr.7: 155507031-155507031 on Build GRCh38
rs751578036 Chr.7: 155506998-155506998 on Build GRCh38
rs775030259 Chr.7: 155507137-155507137 on Build GRCh38
rs772587364 Chr.7: 155507049-155507049 on Build GRCh38
rs147864277 Chr.7: 155507085-155507085 on Build GRCh38
rs763554279 Chr.7: 155507107-155507107 on Build GRCh38
rs771404172 Chr.7: 155507069-155507069 on Build GRCh38
rs1034227914 Chr.7: 155507138-155507138 on Build GRCh38
rs967510923 Chr.7: 155507163-155507163 on Build GRCh38
rs775028734 Chr.7: 155507101-155507101 on Build GRCh38
rs752889931 Chr.7: 155507165-155507165 on Build GRCh38
rs750232079 Chr.7: 155507015-155507015 on Build GRCh38
rs1014848927 Chr.7: 155507034-155507034 on Build GRCh38
rs575349964 Chr.7: 155507171-155507171 on Build GRCh38
rs759814781 Chr.7: 155507084-155507084 on Build GRCh38
rs557017418 Chr.7: 155507167-155507167 on Build GRCh38
rs757231178 Chr.7: 155506999-155506999 on Build GRCh38
rs763611061 Chr.7: 155506993-155506993 on Build GRCh38
rs772062852 Chr.7: 155507051-155507051 on Build GRCh38
rs373619692 Chr.7: 155507068-155507068 on Build GRCh38
rs779845856 Chr.7: 155507158-155507158 on Build GRCh38
rs781183204 Chr.7: 155507004-155507004 on Build GRCh38
rs769250315 Chr.7: 155507062-155507062 on Build GRCh38
rs752318250 Chr.7: 155506991-155506991 on Build GRCh38
rs1021462532 Chr.7: 155507162-155507162 on Build GRCh38
rs777788821 Chr.7: 155507052-155507052 on Build GRCh38
rs144666756 Chr.7: 155506992-155506992 on Build GRCh38
rs10260547 Chr.7: 155506980-155506980 on Build GRCh38
rs748621401 Chr.7: 155507048-155507051 on Build GRCh38
rs773761662 Chr.7: 155507169-155507172 on Build GRCh38
rs748341430 Chr.7: 155507048-155507048 on Build GRCh38
rs376710673 Chr.7: 155507077-155507077 on Build GRCh38
rs535482078 Chr.7: 155507151-155507151 on Build GRCh38
rs755878413 Chr.7: 155507156-155507156 on Build GRCh38
rs753985411 Chr.7: 155506999-155507000 on Build GRCh38
rs896324885 Chr.7: 155506976-155506976 on Build GRCh38
rs1036247369 Chr.7: 155506960-155506960 on Build GRCh38

View Full Product Details