Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 67493919-67494176 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CENPC
Entrez Gene ID
1060
Gene Name
centromere protein C
Gene Aliases
CENP-C, CENPC1, MIF2, hcp-4
UniGene
Hs.479867
Gene Chromosome Location
Chr.4: 67468762-67545601 on Build GRCh38
Transcripts
NM_001812.2 XM_006714064.2 XM_011531542.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1056694 c.2227A>T p.T743S Chr.4: 67493947-67493947 on Build GRCh38
1056692 c.2231G>A p.R744H Chr.4: 67493943-67493943 on Build GRCh38
4598535 c.2254C>T p.R752* Chr.4: 67493920-67493920 on Build GRCh38
1248067 c.2209G>A p.V737I Chr.4: 67493965-67493965 on Build GRCh38
6301738 c.2206A>G p.N736D Chr.4: 67493968-67493968 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs888284594 Chr.4: 67494139-67494139 on Build GRCh38
rs954140297 Chr.4: 67494042-67494042 on Build GRCh38
rs768395203 Chr.4: 67494007-67494007 on Build GRCh38
rs755558750 Chr.4: 67493972-67493972 on Build GRCh38
rs773219449 Chr.4: 67494021-67494021 on Build GRCh38
rs945092359 Chr.4: 67494071-67494071 on Build GRCh38
rs1049519323 Chr.4: 67494138-67494138 on Build GRCh38
rs918838786 Chr.4: 67494096-67494096 on Build GRCh38
rs1019720843 Chr.4: 67494069-67494069 on Build GRCh38
rs965966108 Chr.4: 67494076-67494076 on Build GRCh38
rs372886452 Chr.4: 67493948-67493948 on Build GRCh38
rs989264630 Chr.4: 67494061-67494061 on Build GRCh38
rs748888225 Chr.4: 67494001-67494001 on Build GRCh38
rs747926355 Chr.4: 67494011-67494011 on Build GRCh38
rs779593087 Chr.4: 67493998-67493998 on Build GRCh38
rs369333716 Chr.4: 67493943-67493943 on Build GRCh38
rs750931543 Chr.4: 67493961-67493961 on Build GRCh38
rs552267308 Chr.4: 67494109-67494109 on Build GRCh38
rs936367747 Chr.4: 67494054-67494054 on Build GRCh38
rs540030757 Chr.4: 67494050-67494050 on Build GRCh38
rs1001276262 Chr.4: 67493973-67493973 on Build GRCh38
rs751999791 Chr.4: 67493944-67493944 on Build GRCh38
rs187437348 Chr.4: 67493954-67493954 on Build GRCh38
rs369923101 Chr.4: 67494089-67494089 on Build GRCh38
rs902478793 Chr.4: 67493970-67493970 on Build GRCh38
rs1020655362 Chr.4: 67493962-67493962 on Build GRCh38
rs375834191 Chr.4: 67494020-67494020 on Build GRCh38
rs532675879 Chr.4: 67494009-67494009 on Build GRCh38

View Full Product Details